Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57637602-57637873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7378786 c.968A>G p.H323R Chr.15: 57637729-57637729 on Build GRCh38
3887249 c.1009G>A p.E337K Chr.15: 57637770-57637770 on Build GRCh38
3502717 c.895G>A p.E299K Chr.15: 57637749-57637749 on Build GRCh38
3502714 c.865C>T p.L289F Chr.15: 57637719-57637719 on Build GRCh38
5646257 c.841G>A p.E281K Chr.15: 57637695-57637695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768883780 Chr.15: 57637747-57637747 on Build GRCh38
rs777903156 Chr.15: 57637720-57637720 on Build GRCh38
rs780586287 Chr.15: 57637706-57637706 on Build GRCh38
rs544126764 Chr.15: 57637626-57637626 on Build GRCh38
rs547980794 Chr.15: 57637794-57637794 on Build GRCh38
rs749255427 Chr.15: 57637816-57637816 on Build GRCh38
rs966744343 Chr.15: 57637685-57637685 on Build GRCh38
rs376724574 Chr.15: 57637644-57637644 on Build GRCh38
rs916804857 Chr.15: 57637752-57637752 on Build GRCh38
rs375478526 Chr.15: 57637663-57637663 on Build GRCh38
rs117361082 Chr.15: 57637771-57637771 on Build GRCh38
rs751899628 Chr.15: 57637786-57637786 on Build GRCh38
rs752372480 Chr.15: 57637800-57637800 on Build GRCh38
rs745716943 Chr.15: 57637824-57637824 on Build GRCh38
rs780460164 Chr.15: 57637695-57637695 on Build GRCh38
rs183283064 Chr.15: 57637781-57637781 on Build GRCh38
rs202183202 Chr.15: 57637760-57637760 on Build GRCh38
rs760830089 Chr.15: 57637733-57637733 on Build GRCh38
rs751176172 Chr.15: 57637698-57637698 on Build GRCh38
rs770857203 Chr.15: 57637821-57637821 on Build GRCh38
rs750908979 Chr.15: 57637692-57637692 on Build GRCh38
rs1007125859 Chr.15: 57637823-57637823 on Build GRCh38
rs375528014 Chr.15: 57637812-57637812 on Build GRCh38
rs200107926 Chr.15: 57637659-57637659 on Build GRCh38
rs35202731 Chr.15: 57637745-57637746 on Build GRCh38
rs774141969 Chr.15: 57637730-57637730 on Build GRCh38
rs766665230 Chr.15: 57637782-57637782 on Build GRCh38
rs754614130 Chr.15: 57637793-57637793 on Build GRCh38
rs753713845 Chr.15: 57637815-57637815 on Build GRCh38
rs747617490 Chr.15: 57637718-57637718 on Build GRCh38
rs893027336 Chr.15: 57637839-57637839 on Build GRCh38
rs865864742 Chr.15: 57637770-57637770 on Build GRCh38
rs758820044 Chr.15: 57637693-57637693 on Build GRCh38
rs369878624 Chr.15: 57637822-57637822 on Build GRCh38
rs765286776 Chr.15: 57637762-57637762 on Build GRCh38
rs978161897 Chr.15: 57637688-57637688 on Build GRCh38
rs764097567 Chr.15: 57637655-57637655 on Build GRCh38
rs776805672 Chr.15: 57637750-57637750 on Build GRCh38
rs140229874 Chr.15: 57637729-57637729 on Build GRCh38
rs1018138722 Chr.15: 57637838-57637838 on Build GRCh38
rs754600804 Chr.15: 57637699-57637699 on Build GRCh38

View Full Product Details