Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120402731-120402996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL1
Entrez Gene ID
11167
Gene Name
follistatin like 1
Gene Aliases
FRP, FSL1, MIR198, OCC-1, OCC1, tsc36
UniGene
Hs.269512
Gene Chromosome Location
Chr.3: 120394214-120451071 on Build GRCh38
Transcripts
NM_007085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173888 c.695-1G>C p.? Chr.3: 120402919-120402919 on Build GRCh38
1642058 c.697T>A p.C233S Chr.3: 120402916-120402916 on Build GRCh38
7134554 c.804C>T p.D268D Chr.3: 120402809-120402809 on Build GRCh38
3585825 c.768C>T p.A256A Chr.3: 120402845-120402845 on Build GRCh38
7283660 c.755G>A p.R252H Chr.3: 120402858-120402858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754750214 Chr.3: 120402874-120402874 on Build GRCh38
rs748273138 Chr.3: 120402863-120402863 on Build GRCh38
rs770968995 Chr.3: 120402808-120402808 on Build GRCh38
rs1001119703 Chr.3: 120402852-120402852 on Build GRCh38
rs369840425 Chr.3: 120402818-120402818 on Build GRCh38
rs765824738 Chr.3: 120402937-120402937 on Build GRCh38
rs774017361 Chr.3: 120402821-120402821 on Build GRCh38
rs961544613 Chr.3: 120402884-120402884 on Build GRCh38
rs771983220 Chr.3: 120402936-120402936 on Build GRCh38
rs761595702 Chr.3: 120402837-120402837 on Build GRCh38
rs777989598 Chr.3: 120402792-120402792 on Build GRCh38
rs945802670 Chr.3: 120402799-120402799 on Build GRCh38
rs750011089 Chr.3: 120402858-120402858 on Build GRCh38
rs770056118 Chr.3: 120402917-120402917 on Build GRCh38
rs773003139 Chr.3: 120402931-120402931 on Build GRCh38
rs757536872 Chr.3: 120402876-120402876 on Build GRCh38
rs143843913 Chr.3: 120402897-120402897 on Build GRCh38
rs201866874 Chr.3: 120402875-120402875 on Build GRCh38
rs895437377 Chr.3: 120402867-120402867 on Build GRCh38
rs781248045 Chr.3: 120402882-120402882 on Build GRCh38
rs141738233 Chr.3: 120402893-120402893 on Build GRCh38
rs753732808 Chr.3: 120402772-120402772 on Build GRCh38
rs776588274 Chr.3: 120402945-120402945 on Build GRCh38
rs139355917 Chr.3: 120402809-120402809 on Build GRCh38
rs775865393 Chr.3: 120402816-120402816 on Build GRCh38
rs774625898 Chr.3: 120402910-120402910 on Build GRCh38
rs769875437 Chr.3: 120402885-120402885 on Build GRCh38
rs775694282 Chr.3: 120402956-120402956 on Build GRCh38
rs540641061 Chr.3: 120402854-120402854 on Build GRCh38
rs190114446 Chr.3: 120402778-120402778 on Build GRCh38
rs763320508 Chr.3: 120402817-120402817 on Build GRCh38
rs193167966 Chr.3: 120402967-120402967 on Build GRCh38
rs745944042 Chr.3: 120402868-120402868 on Build GRCh38
rs747216275 Chr.3: 120402800-120402800 on Build GRCh38
rs770357446 Chr.3: 120402812-120402812 on Build GRCh38
rs771916664 Chr.3: 120402921-120402921 on Build GRCh38
rs746365386 Chr.3: 120402810-120402810 on Build GRCh38
rs573306533 Chr.3: 120402887-120402887 on Build GRCh38
rs753729548 Chr.3: 120402871-120402871 on Build GRCh38
rs144093733 Chr.3: 120402896-120402896 on Build GRCh38
rs906888719 Chr.3: 120402832-120402832 on Build GRCh38
rs760685380 Chr.3: 120402859-120402859 on Build GRCh38
rs759443163 Chr.3: 120402949-120402949 on Build GRCh38
rs887101874 Chr.3: 120402788-120402788 on Build GRCh38
rs745885697 Chr.3: 120402883-120402883 on Build GRCh38
rs911210830 Chr.3: 120402904-120402904 on Build GRCh38
rs767668404 Chr.3: 120402961-120402961 on Build GRCh38
rs765071121 Chr.3: 120402953-120402953 on Build GRCh38
rs146464696 Chr.3: 120402954-120402954 on Build GRCh38
rs201107663 Chr.3: 120402928-120402928 on Build GRCh38

View Full Product Details