Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43776009-43776248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL3
Entrez Gene ID
80237
Gene Name
elongation factor for RNA polymerase II 3
Gene Aliases
-
UniGene
Hs.706346
Gene Chromosome Location
Chr.15: 43772600-43777304 on Build GRCh38
Transcripts
NM_025165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299947 c.169T>C p.Y57H Chr.15: 43776151-43776151 on Build GRCh38
6279819 c.260T>A p.L87H Chr.15: 43776060-43776060 on Build GRCh38
6653333 c.196T>C p.S66P Chr.15: 43776124-43776124 on Build GRCh38
1323186 c.218T>C p.V73A Chr.15: 43776102-43776102 on Build GRCh38
4482364 c.259C>T p.L87F Chr.15: 43776061-43776061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756386373 Chr.15: 43776198-43776198 on Build GRCh38
rs772490233 Chr.15: 43776104-43776105 on Build GRCh38
rs764778001 Chr.15: 43776066-43776066 on Build GRCh38
rs955048621 Chr.15: 43776197-43776197 on Build GRCh38
rs867727933 Chr.15: 43776123-43776123 on Build GRCh38
rs745924343 Chr.15: 43776214-43776214 on Build GRCh38
rs751949796 Chr.15: 43776091-43776091 on Build GRCh38
rs111431125 Chr.15: 43776030-43776030 on Build GRCh38
rs78513225 Chr.15: 43776154-43776154 on Build GRCh38
rs760165041 Chr.15: 43776170-43776171 on Build GRCh38
rs573886443 Chr.15: 43776076-43776076 on Build GRCh38
rs781604475 Chr.15: 43776106-43776106 on Build GRCh38
rs772918291 Chr.15: 43776224-43776224 on Build GRCh38
rs542281975 Chr.15: 43776086-43776086 on Build GRCh38
rs768705102 Chr.15: 43776157-43776157 on Build GRCh38
rs753269162 Chr.15: 43776170-43776170 on Build GRCh38
rs767987742 Chr.15: 43776192-43776192 on Build GRCh38
rs899401923 Chr.15: 43776155-43776155 on Build GRCh38
rs569865330 Chr.15: 43776217-43776217 on Build GRCh38
rs775502833 Chr.15: 43776153-43776153 on Build GRCh38
rs771134985 Chr.15: 43776038-43776038 on Build GRCh38
rs765736640 Chr.15: 43776179-43776179 on Build GRCh38
rs764970727 Chr.15: 43776171-43776171 on Build GRCh38
rs772689242 Chr.15: 43776173-43776173 on Build GRCh38
rs963664328 Chr.15: 43776194-43776194 on Build GRCh38
rs768756325 Chr.15: 43776216-43776216 on Build GRCh38
rs776619366 Chr.15: 43776169-43776169 on Build GRCh38
rs767979395 Chr.15: 43776153-43776153 on Build GRCh38
rs755340175 Chr.15: 43776093-43776093 on Build GRCh38
rs747977070 Chr.15: 43776219-43776219 on Build GRCh38
rs767299656 Chr.15: 43776074-43776074 on Build GRCh38
rs781351075 Chr.15: 43776087-43776087 on Build GRCh38
rs774811566 Chr.15: 43776045-43776051 on Build GRCh38
rs757803957 Chr.15: 43776077-43776077 on Build GRCh38
rs147129198 Chr.15: 43776112-43776112 on Build GRCh38
rs751064634 Chr.15: 43776182-43776182 on Build GRCh38
rs763250234 Chr.15: 43776029-43776030 on Build GRCh38
rs183272065 Chr.15: 43776206-43776206 on Build GRCh38
rs755536235 Chr.15: 43776187-43776187 on Build GRCh38
rs770858751 Chr.15: 43776149-43776149 on Build GRCh38
rs564435843 Chr.15: 43776177-43776177 on Build GRCh38
rs749404488 Chr.15: 43776209-43776209 on Build GRCh38
rs533484125 Chr.15: 43776193-43776193 on Build GRCh38
rs759864436 Chr.15: 43776109-43776111 on Build GRCh38
rs541095065 Chr.15: 43776164-43776164 on Build GRCh38
rs760560529 Chr.15: 43776033-43776033 on Build GRCh38
rs753685198 Chr.15: 43776054-43776054 on Build GRCh38
rs778921131 Chr.15: 43776213-43776213 on Build GRCh38
rs143421902 Chr.15: 43776059-43776059 on Build GRCh38
rs148701234 Chr.15: 43776135-43776135 on Build GRCh38
rs766927348 Chr.15: 43776084-43776084 on Build GRCh38
rs761673078 Chr.15: 43776170-43776170 on Build GRCh38
rs201419814 Chr.15: 43776104-43776104 on Build GRCh38
rs756376497 Chr.15: 43776132-43776132 on Build GRCh38
rs763915130 Chr.15: 43776048-43776048 on Build GRCh38
rs769413709 Chr.15: 43776221-43776221 on Build GRCh38
rs1026436404 Chr.15: 43776199-43776199 on Build GRCh38
rs200980642 Chr.15: 43776148-43776148 on Build GRCh38
rs887183846 Chr.15: 43776070-43776070 on Build GRCh38

View Full Product Details