Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206648782-206649053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK3
Entrez Gene ID
8444
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Gene Aliases
DYRK5, RED, REDK, hYAK3-2
UniGene
Hs.164267
Gene Chromosome Location
Chr.1: 206635536-206655158 on Build GRCh38
Transcripts
NM_001004023.1 NM_003582.2 XM_005273315.4 XM_011510061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463839 c.1679G>T p.G560V Chr.1: 206648877-206648877 on Build GRCh38
6991130 c.1624A>G p.I542V Chr.1: 206648927-206648927 on Build GRCh38
463841 c.1734C>T p.P578P Chr.1: 206648932-206648932 on Build GRCh38
1668425 c.1670C>T p.P557L Chr.1: 206648868-206648868 on Build GRCh38
463840 c.1574G>T p.G525V Chr.1: 206648877-206648877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782370025 Chr.1: 206648944-206648944 on Build GRCh38
rs200713840 Chr.1: 206648816-206648816 on Build GRCh38
rs782111789 Chr.1: 206648998-206648998 on Build GRCh38
rs201145019 Chr.1: 206648991-206648991 on Build GRCh38
rs948676451 Chr.1: 206648833-206648833 on Build GRCh38
rs1029374889 Chr.1: 206648997-206648997 on Build GRCh38
rs201787773 Chr.1: 206649026-206649026 on Build GRCh38
rs200820369 Chr.1: 206648806-206648806 on Build GRCh38
rs568923051 Chr.1: 206648905-206648905 on Build GRCh38
rs781787559 Chr.1: 206648887-206648887 on Build GRCh38
rs781963697 Chr.1: 206648863-206648863 on Build GRCh38
rs529521269 Chr.1: 206648847-206648847 on Build GRCh38
rs201711218 Chr.1: 206648817-206648817 on Build GRCh38
rs137901718 Chr.1: 206648844-206648844 on Build GRCh38
rs199976942 Chr.1: 206648808-206648808 on Build GRCh38
rs782636886 Chr.1: 206648815-206648815 on Build GRCh38
rs782094487 Chr.1: 206648865-206648865 on Build GRCh38
rs969312127 Chr.1: 206649032-206649032 on Build GRCh38
rs782047095 Chr.1: 206648988-206648988 on Build GRCh38
rs1005365600 Chr.1: 206648959-206648959 on Build GRCh38
rs782754292 Chr.1: 206648867-206648867 on Build GRCh38
rs782292094 Chr.1: 206648913-206648913 on Build GRCh38
rs199768627 Chr.1: 206648809-206648809 on Build GRCh38
rs782598896 Chr.1: 206648942-206648942 on Build GRCh38
rs781860367 Chr.1: 206648893-206648893 on Build GRCh38
rs782601735 Chr.1: 206648891-206648891 on Build GRCh38
rs782780113 Chr.1: 206648999-206648999 on Build GRCh38
rs781821673 Chr.1: 206648993-206648993 on Build GRCh38
rs782361196 Chr.1: 206648814-206648814 on Build GRCh38
rs782138320 Chr.1: 206648877-206648877 on Build GRCh38
rs781820020 Chr.1: 206648976-206648976 on Build GRCh38
rs200134906 Chr.1: 206649005-206649005 on Build GRCh38
rs782055369 Chr.1: 206648843-206648843 on Build GRCh38
rs782036572 Chr.1: 206648836-206648836 on Build GRCh38
rs200892244 Chr.1: 206648886-206648886 on Build GRCh38
rs781981992 Chr.1: 206648965-206648965 on Build GRCh38
rs199879907 Chr.1: 206648873-206648873 on Build GRCh38
rs782033949 Chr.1: 206648983-206648983 on Build GRCh38
rs200225267 Chr.1: 206649027-206649027 on Build GRCh38
rs551854696 Chr.1: 206648976-206648976 on Build GRCh38
rs782671806 Chr.1: 206648907-206648907 on Build GRCh38
rs148139981 Chr.1: 206648978-206648978 on Build GRCh38
rs192975758 Chr.1: 206649003-206649003 on Build GRCh38
rs782216378 Chr.1: 206648943-206648943 on Build GRCh38
rs141266804 Chr.1: 206648919-206648919 on Build GRCh38
rs74147411 Chr.1: 206649013-206649013 on Build GRCh38
rs549234649 Chr.1: 206648890-206648890 on Build GRCh38

View Full Product Details