Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49588405-49588678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM186B
Entrez Gene ID
84070
Gene Name
family with sequence similarity 186 member B
Gene Aliases
C12orf25
UniGene
Hs.524406
Gene Chromosome Location
Chr.12: 49582875-49605710 on Build GRCh38
Transcripts
NM_032130.2 NR_027450.1 XM_006719625.2 XM_006719626.2 XM_006719627.3 XM_011538796.2 XM_011538797.2 XM_017020008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6214387 c.2140T>G p.S714A Chr.12: 49588578-49588578 on Build GRCh38
4906018 c.2191C>T p.R731C Chr.12: 49588527-49588527 on Build GRCh38
7367220 c.2452C>T p.R818W Chr.12: 49588536-49588536 on Build GRCh38
3461694 c.2461C>T p.R821C Chr.12: 49588527-49588527 on Build GRCh38
5503903 c.2466C>A p.P822P Chr.12: 49588522-49588522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753802239 Chr.12: 49588495-49588495 on Build GRCh38
rs76224014 Chr.12: 49588455-49588455 on Build GRCh38
rs370627855 Chr.12: 49588564-49588564 on Build GRCh38
rs551256193 Chr.12: 49588643-49588643 on Build GRCh38
rs759324336 Chr.12: 49588585-49588585 on Build GRCh38
rs764246617 Chr.12: 49588601-49588601 on Build GRCh38
rs770833500 Chr.12: 49588576-49588576 on Build GRCh38
rs375978412 Chr.12: 49588541-49588541 on Build GRCh38
rs893124968 Chr.12: 49588442-49588442 on Build GRCh38
rs773284841 Chr.12: 49588493-49588496 on Build GRCh38
rs377415942 Chr.12: 49588614-49588614 on Build GRCh38
rs202162733 Chr.12: 49588524-49588524 on Build GRCh38
rs757363870 Chr.12: 49588526-49588526 on Build GRCh38
rs544511372 Chr.12: 49588515-49588515 on Build GRCh38
rs915553222 Chr.12: 49588527-49588527 on Build GRCh38
rs373146376 Chr.12: 49588431-49588431 on Build GRCh38
rs746271849 Chr.12: 49588528-49588528 on Build GRCh38
rs149783745 Chr.12: 49588527-49588527 on Build GRCh38
rs766456589 Chr.12: 49588492-49588492 on Build GRCh38
rs778103645 Chr.12: 49588449-49588449 on Build GRCh38
rs147282399 Chr.12: 49588481-49588481 on Build GRCh38
rs770162543 Chr.12: 49588477-49588477 on Build GRCh38
rs760722128 Chr.12: 49588484-49588484 on Build GRCh38
rs1054596016 Chr.12: 49588612-49588612 on Build GRCh38
rs140789587 Chr.12: 49588482-49588482 on Build GRCh38
rs769861102 Chr.12: 49588467-49588467 on Build GRCh38
rs760593253 Chr.12: 49588575-49588575 on Build GRCh38
rs76361899 Chr.12: 49588559-49588559 on Build GRCh38
rs771367977 Chr.12: 49588595-49588612 on Build GRCh38
rs201014369 Chr.12: 49588460-49588460 on Build GRCh38
rs781467829 Chr.12: 49588450-49588450 on Build GRCh38
rs765425087 Chr.12: 49588436-49588436 on Build GRCh38
rs763459678 Chr.12: 49588502-49588502 on Build GRCh38
rs78249505 Chr.12: 49588536-49588536 on Build GRCh38
rs531081568 Chr.12: 49588639-49588639 on Build GRCh38
rs148973222 Chr.12: 49588459-49588459 on Build GRCh38
rs201521699 Chr.12: 49588443-49588443 on Build GRCh38
rs950298571 Chr.12: 49588579-49588579 on Build GRCh38
rs767776628 Chr.12: 49588636-49588636 on Build GRCh38
rs528638783 Chr.12: 49588538-49588538 on Build GRCh38
rs151051569 Chr.12: 49588535-49588535 on Build GRCh38
rs866301333 Chr.12: 49588485-49588485 on Build GRCh38
rs758561286 Chr.12: 49588507-49588507 on Build GRCh38
rs908119340 Chr.12: 49588570-49588570 on Build GRCh38
rs775908209 Chr.12: 49588605-49588605 on Build GRCh38
rs565488464 Chr.12: 49588584-49588584 on Build GRCh38
rs754386661 Chr.12: 49588646-49588646 on Build GRCh38
rs746387916 Chr.12: 49588452-49588452 on Build GRCh38
rs770245417 Chr.12: 49588454-49588454 on Build GRCh38
rs751544178 Chr.12: 49588607-49588607 on Build GRCh38
rs369883467 Chr.12: 49588433-49588433 on Build GRCh38
rs758376053 Chr.12: 49588444-49588444 on Build GRCh38

View Full Product Details