Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169544668-169544941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD1
Entrez Gene ID
79675
Gene Name
FAST kinase domains 1
Gene Aliases
-
UniGene
Hs.529276
Gene Chromosome Location
Chr.2: 169527879-169573921 on Build GRCh38
Transcripts
NM_001281476.2 NM_001322046.1 NM_001322048.1 NM_001322049.1 NM_024622.5 NR_104020.2 XM_006712750.3 XM_006712751.3 XM_017004911.1 XM_017004912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212212 c.1811A>T p.Y604F Chr.2: 169544726-169544726 on Build GRCh38
4428246 c.1774G>T p.E592* Chr.2: 169544763-169544763 on Build GRCh38
3425437 c.1717A>G p.I573V Chr.2: 169544820-169544820 on Build GRCh38
244419 c.1706A>G p.H569R Chr.2: 169544831-169544831 on Build GRCh38
4773750 c.1800delT p.N602fs*4 Chr.2: 169544737-169544737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748229894 Chr.2: 169544698-169544698 on Build GRCh38
rs973328717 Chr.2: 169544904-169544904 on Build GRCh38
rs1013151191 Chr.2: 169544782-169544782 on Build GRCh38
rs541992657 Chr.2: 169544800-169544800 on Build GRCh38
rs138073323 Chr.2: 169544746-169544746 on Build GRCh38
rs777224591 Chr.2: 169544716-169544716 on Build GRCh38
rs762097839 Chr.2: 169544746-169544747 on Build GRCh38
rs765570133 Chr.2: 169544832-169544832 on Build GRCh38
rs756397050 Chr.2: 169544848-169544848 on Build GRCh38
rs773730794 Chr.2: 169544719-169544719 on Build GRCh38
rs759405753 Chr.2: 169544745-169544745 on Build GRCh38
rs761596946 Chr.2: 169544821-169544821 on Build GRCh38
rs144290701 Chr.2: 169544832-169544832 on Build GRCh38
rs139546135 Chr.2: 169544797-169544797 on Build GRCh38
rs779873542 Chr.2: 169544845-169544845 on Build GRCh38
rs758211801 Chr.2: 169544841-169544841 on Build GRCh38
rs148245019 Chr.2: 169544738-169544738 on Build GRCh38
rs779510022 Chr.2: 169544857-169544857 on Build GRCh38
rs771797962 Chr.2: 169544804-169544804 on Build GRCh38
rs771279778 Chr.2: 169544855-169544855 on Build GRCh38
rs755087519 Chr.2: 169544775-169544775 on Build GRCh38
rs763566011 Chr.2: 169544725-169544725 on Build GRCh38
rs749757992 Chr.2: 169544794-169544794 on Build GRCh38
rs777819670 Chr.2: 169544849-169544849 on Build GRCh38
rs763221593 Chr.2: 169544887-169544888 on Build GRCh38
rs746407002 Chr.2: 169544861-169544861 on Build GRCh38
rs749402081 Chr.2: 169544854-169544854 on Build GRCh38
rs372186353 Chr.2: 169544903-169544903 on Build GRCh38
rs766623965 Chr.2: 169544727-169544727 on Build GRCh38
rs774848429 Chr.2: 169544728-169544728 on Build GRCh38
rs748178523 Chr.2: 169544791-169544791 on Build GRCh38
rs778557116 Chr.2: 169544756-169544756 on Build GRCh38
rs770247137 Chr.2: 169544705-169544705 on Build GRCh38
rs561762592 Chr.2: 169544805-169544805 on Build GRCh38
rs545647875 Chr.2: 169544723-169544723 on Build GRCh38
rs759954911 Chr.2: 169544801-169544801 on Build GRCh38
rs750676911 Chr.2: 169544860-169544860 on Build GRCh38
rs371143972 Chr.2: 169544839-169544839 on Build GRCh38
rs965095567 Chr.2: 169544895-169544895 on Build GRCh38
rs199713276 Chr.2: 169544747-169544747 on Build GRCh38
rs760388786 Chr.2: 169544873-169544873 on Build GRCh38
rs765224579 Chr.2: 169544835-169544835 on Build GRCh38
rs369119259 Chr.2: 169544768-169544768 on Build GRCh38
rs148920574 Chr.2: 169544739-169544739 on Build GRCh38
rs772530384 Chr.2: 169544863-169544863 on Build GRCh38
rs781422946 Chr.2: 169544790-169544790 on Build GRCh38
rs753444362 Chr.2: 169544816-169544816 on Build GRCh38
rs769844413 Chr.2: 169544793-169544793 on Build GRCh38
rs1043730999 Chr.2: 169544785-169544785 on Build GRCh38
rs750603867 Chr.2: 169544910-169544910 on Build GRCh38
rs776061701 Chr.2: 169544870-169544870 on Build GRCh38
rs765567122 Chr.2: 169544902-169544902 on Build GRCh38
rs377405136 Chr.2: 169544778-169544778 on Build GRCh38
rs759099441 Chr.2: 169544733-169544733 on Build GRCh38
rs142046183 Chr.2: 169544737-169544737 on Build GRCh38

View Full Product Details