Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2589249-2589516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213B
Entrez Gene ID
127281
Gene Name
family with sequence similarity 213 member B
Gene Aliases
C1orf93
UniGene
Hs.462033
Gene Chromosome Location
Chr.1: 2586460-2591469 on Build GRCh38
Transcripts
NM_001195736.1 NM_001195737.1 NM_001195738.1 NM_001195740.1 NM_001195741.1 NM_152371.3 NR_036637.1 NR_036638.1 XM_006710354.3 XM_011540664.1 XM_011540665.1 XM_011540666.1

SNP Detail

SNP ID Chromosome Location
rs1011533178 Chr.1: 2589312-2589312 on Build GRCh38
rs761193255 Chr.1: 2589376-2589376 on Build GRCh38
rs751489355 Chr.1: 2589399-2589399 on Build GRCh38
rs748601441 Chr.1: 2589364-2589364 on Build GRCh38
rs554575486 Chr.1: 2589406-2589406 on Build GRCh38
rs761836788 Chr.1: 2589479-2589479 on Build GRCh38
rs890920482 Chr.1: 2589279-2589279 on Build GRCh38
rs969425029 Chr.1: 2589369-2589369 on Build GRCh38
rs998663560 Chr.1: 2589291-2589291 on Build GRCh38
rs960838454 Chr.1: 2589383-2589383 on Build GRCh38
rs1031495295 Chr.1: 2589298-2589298 on Build GRCh38
rs751401388 Chr.1: 2589475-2589475 on Build GRCh38
rs997856343 Chr.1: 2589283-2589283 on Build GRCh38
rs373771434 Chr.1: 2589393-2589393 on Build GRCh38
rs200820865 Chr.1: 2589434-2589434 on Build GRCh38
rs184894716 Chr.1: 2589275-2589275 on Build GRCh38
rs542482574 Chr.1: 2589293-2589293 on Build GRCh38
rs967015202 Chr.1: 2589342-2589342 on Build GRCh38
rs762843452 Chr.1: 2589395-2589395 on Build GRCh38
rs978868857 Chr.1: 2589350-2589350 on Build GRCh38
rs1050808904 Chr.1: 2589278-2589278 on Build GRCh38
rs776109580 Chr.1: 2589450-2589450 on Build GRCh38
rs1022710408 Chr.1: 2589353-2589353 on Build GRCh38
rs763823632 Chr.1: 2589396-2589396 on Build GRCh38
rs370258722 Chr.1: 2589447-2589447 on Build GRCh38
rs377557907 Chr.1: 2589372-2589372 on Build GRCh38
rs759137607 Chr.1: 2589464-2589464 on Build GRCh38
rs778125126 Chr.1: 2589435-2589435 on Build GRCh38
rs754038181 Chr.1: 2589419-2589419 on Build GRCh38
rs920894175 Chr.1: 2589454-2589454 on Build GRCh38
rs770371698 Chr.1: 2589380-2589380 on Build GRCh38
rs780215273 Chr.1: 2589413-2589413 on Build GRCh38
rs199566846 Chr.1: 2589430-2589430 on Build GRCh38
rs771498223 Chr.1: 2589448-2589448 on Build GRCh38
rs149857424 Chr.1: 2589381-2589381 on Build GRCh38
rs759188340 Chr.1: 2589387-2589387 on Build GRCh38
rs762533120 Chr.1: 2589468-2589468 on Build GRCh38
rs932336775 Chr.1: 2589460-2589460 on Build GRCh38
rs764999992 Chr.1: 2589389-2589389 on Build GRCh38
rs371068003 Chr.1: 2589471-2589471 on Build GRCh38
rs779243269 Chr.1: 2589426-2589426 on Build GRCh38
rs375431254 Chr.1: 2589466-2589466 on Build GRCh38
rs78851285 Chr.1: 2589304-2589304 on Build GRCh38
rs769303363 Chr.1: 2589465-2589465 on Build GRCh38
rs763580800 Chr.1: 2589441-2589448 on Build GRCh38
rs762498028 Chr.1: 2589416-2589419 on Build GRCh38
rs773671370 Chr.1: 2589373-2589373 on Build GRCh38

View Full Product Details