Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30778357-30778618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188B
Entrez Gene ID
84182
Gene Name
family with sequence similarity 188 member B
Gene Aliases
C7orf67
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30771417-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Gene Symbol
INMT-FAM188B
Entrez Gene ID
100526825
Gene Name
INMT-FAM188B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30752135-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280973 c.142C>T p.R48* Chr.7: 30778510-30778510 on Build GRCh38
6659138 c.100C>T p.R34C Chr.7: 30778468-30778468 on Build GRCh38
1089097 c.160G>T p.E54* Chr.7: 30778528-30778528 on Build GRCh38
1450358 c.154C>T p.H52Y Chr.7: 30778522-30778522 on Build GRCh38
185137 c.143G>A p.R48Q Chr.7: 30778511-30778511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778312582 Chr.7: 30778575-30778575 on Build GRCh38
rs373430307 Chr.7: 30778409-30778409 on Build GRCh38
rs985282218 Chr.7: 30778599-30778599 on Build GRCh38
rs146665703 Chr.7: 30778400-30778400 on Build GRCh38
rs775572814 Chr.7: 30778523-30778523 on Build GRCh38
rs747484902 Chr.7: 30778511-30778511 on Build GRCh38
rs749950242 Chr.7: 30778550-30778550 on Build GRCh38
rs762589653 Chr.7: 30778467-30778467 on Build GRCh38
rs770060133 Chr.7: 30778516-30778516 on Build GRCh38
rs767179684 Chr.7: 30778522-30778522 on Build GRCh38
rs200053449 Chr.7: 30778512-30778512 on Build GRCh38
rs372698074 Chr.7: 30778567-30778567 on Build GRCh38
rs377520823 Chr.7: 30778475-30778475 on Build GRCh38
rs760095103 Chr.7: 30778554-30778554 on Build GRCh38
rs79584800 Chr.7: 30778585-30778585 on Build GRCh38
rs779370119 Chr.7: 30778408-30778408 on Build GRCh38
rs188045001 Chr.7: 30778474-30778474 on Build GRCh38
rs78628968 Chr.7: 30778581-30778581 on Build GRCh38
rs769751946 Chr.7: 30778589-30778589 on Build GRCh38
rs757852306 Chr.7: 30778580-30778580 on Build GRCh38
rs771111353 Chr.7: 30778446-30778446 on Build GRCh38
rs758873645 Chr.7: 30778568-30778568 on Build GRCh38
rs1015017308 Chr.7: 30778440-30778440 on Build GRCh38
rs548010936 Chr.7: 30778515-30778515 on Build GRCh38
rs778472100 Chr.7: 30778504-30778504 on Build GRCh38
rs767396581 Chr.7: 30778548-30778548 on Build GRCh38
rs761773936 Chr.7: 30778540-30778540 on Build GRCh38
rs868446099 Chr.7: 30778532-30778532 on Build GRCh38
rs750399016 Chr.7: 30778483-30778483 on Build GRCh38
rs1005469117 Chr.7: 30778411-30778411 on Build GRCh38
rs149008261 Chr.7: 30778448-30778448 on Build GRCh38
rs773849633 Chr.7: 30778419-30778419 on Build GRCh38
rs755342881 Chr.7: 30778407-30778407 on Build GRCh38
rs185223356 Chr.7: 30778410-30778410 on Build GRCh38
rs368374263 Chr.7: 30778527-30778527 on Build GRCh38
rs758674640 Chr.7: 30778495-30778495 on Build GRCh38
rs781737529 Chr.7: 30778582-30778582 on Build GRCh38
rs753168544 Chr.7: 30778489-30778489 on Build GRCh38
rs764087591 Chr.7: 30778468-30778468 on Build GRCh38
rs780883742 Chr.7: 30778399-30778399 on Build GRCh38
rs564508518 Chr.7: 30778404-30778404 on Build GRCh38
rs1001543344 Chr.7: 30778405-30778405 on Build GRCh38
rs751922948 Chr.7: 30778579-30778579 on Build GRCh38
rs374974001 Chr.7: 30778456-30778456 on Build GRCh38
rs11762566 Chr.7: 30778382-30778382 on Build GRCh38
rs751487634 Chr.7: 30778469-30778469 on Build GRCh38
rs772684983 Chr.7: 30778591-30778591 on Build GRCh38
rs768420868 Chr.7: 30778535-30778535 on Build GRCh38
rs201946407 Chr.7: 30778526-30778526 on Build GRCh38
rs779502461 Chr.7: 30778484-30778484 on Build GRCh38
rs767437245 Chr.7: 30778446-30778447 on Build GRCh38
rs774314848 Chr.7: 30778537-30778537 on Build GRCh38
rs200855134 Chr.7: 30778383-30778383 on Build GRCh38
rs747141268 Chr.7: 30778423-30778423 on Build GRCh38
rs775370060 Chr.7: 30778461-30778461 on Build GRCh38
rs756756551 Chr.7: 30778384-30778384 on Build GRCh38

View Full Product Details