Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119035902-119036145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3A
Entrez Gene ID
8661
Gene Name
eukaryotic translation initiation factor 3 subunit A
Gene Aliases
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185
UniGene
Hs.523299
Gene Chromosome Location
Chr.10: 119035029-119080822 on Build GRCh38
Transcripts
NM_003750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2055392 c.4097G>A p.R1366H Chr.10: 119036091-119036091 on Build GRCh38
427135 c.4145G>A p.R1382H Chr.10: 119036043-119036043 on Build GRCh38
1251078 c.4142G>A p.R1381Q Chr.10: 119036046-119036046 on Build GRCh38
915308 c.4100G>A p.R1367H Chr.10: 119036088-119036088 on Build GRCh38
3934946 c.4044A>G p.E1348E Chr.10: 119036144-119036144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017028448 Chr.10: 119036016-119036016 on Build GRCh38
rs778671067 Chr.10: 119036103-119036103 on Build GRCh38
rs770087176 Chr.10: 119036009-119036010 on Build GRCh38
rs558214917 Chr.10: 119036013-119036013 on Build GRCh38
rs749107031 Chr.10: 119036038-119036045 on Build GRCh38
rs765705109 Chr.10: 119036044-119036044 on Build GRCh38
rs116931465 Chr.10: 119035994-119035994 on Build GRCh38
rs578051436 Chr.10: 119036074-119036074 on Build GRCh38
rs754701550 Chr.10: 119036097-119036097 on Build GRCh38
rs143903271 Chr.10: 119036091-119036091 on Build GRCh38
rs768516430 Chr.10: 119036038-119036039 on Build GRCh38
rs764426395 Chr.10: 119036088-119036088 on Build GRCh38
rs771820186 Chr.10: 119035990-119035990 on Build GRCh38
rs753786295 Chr.10: 119036092-119036092 on Build GRCh38
rs998426600 Chr.10: 119035934-119035934 on Build GRCh38
rs775993464 Chr.10: 119036054-119036054 on Build GRCh38
rs777303870 Chr.10: 119036014-119036014 on Build GRCh38
rs775208395 Chr.10: 119036034-119036035 on Build GRCh38
rs746575770 Chr.10: 119035998-119035998 on Build GRCh38
rs747759660 Chr.10: 119035988-119035988 on Build GRCh38
rs951348484 Chr.10: 119035976-119035977 on Build GRCh38
rs61729181 Chr.10: 119036114-119036114 on Build GRCh38
rs575905159 Chr.10: 119035937-119035937 on Build GRCh38
rs759924980 Chr.10: 119036034-119036034 on Build GRCh38
rs751564493 Chr.10: 119036089-119036089 on Build GRCh38

View Full Product Details