Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225831734-225832007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX1
Entrez Gene ID
2052
Gene Name
epoxide hydrolase 1
Gene Aliases
EPHX, EPOX, HYL1, MEH
UniGene
Hs.89649
Gene Chromosome Location
Chr.1: 225810074-225845563 on Build GRCh38
Transcripts
NM_000120.3 NM_001136018.3 NM_001291163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3484220 c.210C>T p.F70F Chr.1: 225831805-225831805 on Build GRCh38
4232612 c.211C>T p.R71C Chr.1: 225831806-225831806 on Build GRCh38
1689916 c.340C>T p.P114S Chr.1: 225831935-225831935 on Build GRCh38
3484219 c.201C>T p.I67I Chr.1: 225831796-225831796 on Build GRCh38
4421580 c.364+1G>A p.? Chr.1: 225831960-225831960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752029504 Chr.1: 225831809-225831809 on Build GRCh38
rs750062692 Chr.1: 225831796-225831796 on Build GRCh38
rs372841024 Chr.1: 225831869-225831869 on Build GRCh38
rs773208265 Chr.1: 225831947-225831947 on Build GRCh38
rs58623835 Chr.1: 225831807-225831807 on Build GRCh38
rs781728701 Chr.1: 225831903-225831903 on Build GRCh38
rs376089070 Chr.1: 225831975-225831975 on Build GRCh38
rs767269084 Chr.1: 225831791-225831791 on Build GRCh38
rs1051740 Chr.1: 225831932-225831932 on Build GRCh38
rs760314091 Chr.1: 225831797-225831797 on Build GRCh38
rs758998279 Chr.1: 225831887-225831887 on Build GRCh38
rs922436327 Chr.1: 225831863-225831863 on Build GRCh38
rs756575376 Chr.1: 225831908-225831908 on Build GRCh38
rs1131873 Chr.1: 225831952-225831952 on Build GRCh38
rs933413426 Chr.1: 225831875-225831875 on Build GRCh38
rs957684939 Chr.1: 225831817-225831817 on Build GRCh38
rs781476194 Chr.1: 225831815-225831815 on Build GRCh38
rs761701159 Chr.1: 225831856-225831856 on Build GRCh38
rs145942697 Chr.1: 225831841-225831841 on Build GRCh38
rs759251392 Chr.1: 225831976-225831976 on Build GRCh38
rs145813024 Chr.1: 225831930-225831930 on Build GRCh38
rs770735746 Chr.1: 225831974-225831974 on Build GRCh38
rs148240980 Chr.1: 225831806-225831806 on Build GRCh38
rs759135765 Chr.1: 225831780-225831781 on Build GRCh38
rs55798709 Chr.1: 225831826-225831826 on Build GRCh38
rs753521411 Chr.1: 225831882-225831882 on Build GRCh38
rs768876566 Chr.1: 225831844-225831844 on Build GRCh38
rs774319268 Chr.1: 225831853-225831853 on Build GRCh38
rs772942082 Chr.1: 225831860-225831860 on Build GRCh38
rs200804772 Chr.1: 225831914-225831914 on Build GRCh38
rs1037939277 Chr.1: 225831762-225831762 on Build GRCh38
rs750685152 Chr.1: 225831907-225831907 on Build GRCh38
rs146362595 Chr.1: 225831785-225831785 on Build GRCh38
rs770081240 Chr.1: 225831827-225831827 on Build GRCh38
rs372504987 Chr.1: 225831763-225831763 on Build GRCh38
rs764776530 Chr.1: 225831888-225831888 on Build GRCh38
rs757610140 Chr.1: 225831811-225831811 on Build GRCh38
rs767066517 Chr.1: 225831969-225831970 on Build GRCh38
rs942988538 Chr.1: 225831953-225831953 on Build GRCh38
rs1039848704 Chr.1: 225831787-225831787 on Build GRCh38
rs755081785 Chr.1: 225831916-225831916 on Build GRCh38
rs67892231 Chr.1: 225831945-225831945 on Build GRCh38
rs2234699 Chr.1: 225831898-225831898 on Build GRCh38
rs750348399 Chr.1: 225831928-225831928 on Build GRCh38
rs780371051 Chr.1: 225831913-225831913 on Build GRCh38
rs751180258 Chr.1: 225831855-225831855 on Build GRCh38
rs4986931 Chr.1: 225831885-225831885 on Build GRCh38
rs765952051 Chr.1: 225831880-225831880 on Build GRCh38
rs139835798 Chr.1: 225831859-225831859 on Build GRCh38
rs192093363 Chr.1: 225831758-225831758 on Build GRCh38
rs760479841 Chr.1: 225831868-225831868 on Build GRCh38
rs778213283 Chr.1: 225831808-225831808 on Build GRCh38
rs559949219 Chr.1: 225831760-225831760 on Build GRCh38
rs780874102 Chr.1: 225831899-225831899 on Build GRCh38
rs1131873 Chr.1: 225831952-225831952 on Build GRCh38
rs1007482363 Chr.1: 225831779-225831779 on Build GRCh38
rs369096677 Chr.1: 225831830-225831830 on Build GRCh38
rs2234698 Chr.1: 225831799-225831799 on Build GRCh38
rs768645557 Chr.1: 225831770-225831770 on Build GRCh38
rs752101788 Chr.1: 225831891-225831891 on Build GRCh38
rs942003770 Chr.1: 225831921-225831921 on Build GRCh38

View Full Product Details