Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17874152-17874425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGL1
Entrez Gene ID
2267
Gene Name
fibrinogen like 1
Gene Aliases
HFREP1, HP-041, LFIRE-1, LFIRE1
UniGene
Hs.491143
Gene Chromosome Location
Chr.8: 17864380-17909983 on Build GRCh38
Transcripts
NM_004467.3 NM_147203.2 NM_201552.1 NM_201553.1 XM_017013232.1 XM_017013233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569127 c.390T>A p.S130R Chr.8: 17874376-17874376 on Build GRCh38
4684640 c.397T>G p.F133V Chr.8: 17874369-17874369 on Build GRCh38
5409146 c.391G>A p.E131K Chr.8: 17874375-17874375 on Build GRCh38
6663866 c.346G>A p.D116N Chr.8: 17874420-17874420 on Build GRCh38
1097595 c.345C>T p.S115S Chr.8: 17874421-17874421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759329624 Chr.8: 17874325-17874325 on Build GRCh38
rs1024220083 Chr.8: 17874331-17874331 on Build GRCh38
rs755328401 Chr.8: 17874364-17874364 on Build GRCh38
rs753764266 Chr.8: 17874343-17874343 on Build GRCh38
rs747323904 Chr.8: 17874362-17874362 on Build GRCh38
rs982054830 Chr.8: 17874347-17874347 on Build GRCh38
rs760572785 Chr.8: 17874339-17874339 on Build GRCh38
rs764055375 Chr.8: 17874341-17874341 on Build GRCh38
rs750635580 Chr.8: 17874352-17874352 on Build GRCh38
rs770470957 Chr.8: 17874372-17874372 on Build GRCh38
rs775315817 Chr.8: 17874382-17874382 on Build GRCh38
rs921112442 Chr.8: 17874206-17874206 on Build GRCh38
rs1012735089 Chr.8: 17874286-17874286 on Build GRCh38
rs988226157 Chr.8: 17874363-17874363 on Build GRCh38
rs765287617 Chr.8: 17874346-17874346 on Build GRCh38
rs367543613 Chr.8: 17874328-17874328 on Build GRCh38
rs113038731 Chr.8: 17874361-17874361 on Build GRCh38
rs768389622 Chr.8: 17874389-17874389 on Build GRCh38
rs573104574 Chr.8: 17874248-17874248 on Build GRCh38
rs374709305 Chr.8: 17874345-17874345 on Build GRCh38
rs112170258 Chr.8: 17874360-17874360 on Build GRCh38
rs1050663755 Chr.8: 17874176-17874176 on Build GRCh38
rs751525129 Chr.8: 17874175-17874175 on Build GRCh38
rs778636832 Chr.8: 17874366-17874367 on Build GRCh38
rs200686011 Chr.8: 17874359-17874359 on Build GRCh38
rs188608946 Chr.8: 17874184-17874184 on Build GRCh38
rs771804269 Chr.8: 17874381-17874381 on Build GRCh38
rs995102614 Chr.8: 17874302-17874302 on Build GRCh38
rs946666759 Chr.8: 17874190-17874190 on Build GRCh38
rs748350363 Chr.8: 17874315-17874315 on Build GRCh38
rs968333930 Chr.8: 17874398-17874398 on Build GRCh38
rs114704900 Chr.8: 17874279-17874279 on Build GRCh38
rs1022801063 Chr.8: 17874310-17874310 on Build GRCh38
rs34871936 Chr.8: 17874367-17874367 on Build GRCh38
rs886761909 Chr.8: 17874253-17874253 on Build GRCh38
rs1008535991 Chr.8: 17874197-17874197 on Build GRCh38
rs774184250 Chr.8: 17874213-17874213 on Build GRCh38
rs993276081 Chr.8: 17874233-17874233 on Build GRCh38
rs576102008 Chr.8: 17874221-17874221 on Build GRCh38
rs111376859 Chr.8: 17874230-17874230 on Build GRCh38
rs748791444 Chr.8: 17874313-17874313 on Build GRCh38
rs760490854 Chr.8: 17874388-17874388 on Build GRCh38
rs899384615 Chr.8: 17874290-17874290 on Build GRCh38
rs34239530 Chr.8: 17874405-17874405 on Build GRCh38
rs61744652 Chr.8: 17874390-17874390 on Build GRCh38
rs199781222 Chr.8: 17874379-17874379 on Build GRCh38
rs988055954 Chr.8: 17874174-17874176 on Build GRCh38
rs533463660 Chr.8: 17874402-17874402 on Build GRCh38
rs754874876 Chr.8: 17874364-17874369 on Build GRCh38
rs578014942 Chr.8: 17874173-17874173 on Build GRCh38
rs748661649 Chr.8: 17874370-17874370 on Build GRCh38
rs1035089432 Chr.8: 17874375-17874375 on Build GRCh38
rs368503160 Chr.8: 17874354-17874354 on Build GRCh38
rs372364435 Chr.8: 17874376-17874376 on Build GRCh38
rs370532467 Chr.8: 17874330-17874330 on Build GRCh38
rs933726703 Chr.8: 17874207-17874207 on Build GRCh38
rs765222489 Chr.8: 17874392-17874392 on Build GRCh38
rs116032579 Chr.8: 17874198-17874198 on Build GRCh38

View Full Product Details