Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143868621-143868864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs782810532 Chr.8: 143868680-143868680 on Build GRCh38
rs782760991 Chr.8: 143868803-143868803 on Build GRCh38
rs782537971 Chr.8: 143868724-143868724 on Build GRCh38
rs368668109 Chr.8: 143868817-143868817 on Build GRCh38
rs983173970 Chr.8: 143868791-143868791 on Build GRCh38
rs782557877 Chr.8: 143868814-143868814 on Build GRCh38
rs781841573 Chr.8: 143868693-143868693 on Build GRCh38
rs375222059 Chr.8: 143868838-143868838 on Build GRCh38
rs201611695 Chr.8: 143868689-143868698 on Build GRCh38
rs782309931 Chr.8: 143868762-143868762 on Build GRCh38
rs533996119 Chr.8: 143868829-143868829 on Build GRCh38
rs927766632 Chr.8: 143868657-143868657 on Build GRCh38
rs373092735 Chr.8: 143868830-143868830 on Build GRCh38
rs368146843 Chr.8: 143868705-143868705 on Build GRCh38
rs782009256 Chr.8: 143868651-143868651 on Build GRCh38
rs1017745299 Chr.8: 143868758-143868758 on Build GRCh38
rs781881083 Chr.8: 143868717-143868717 on Build GRCh38
rs782814100 Chr.8: 143868832-143868832 on Build GRCh38
rs782728846 Chr.8: 143868692-143868692 on Build GRCh38
rs6558399 Chr.8: 143868735-143868735 on Build GRCh38
rs201602383 Chr.8: 143868822-143868822 on Build GRCh38
rs782805448 Chr.8: 143868773-143868773 on Build GRCh38
rs782101178 Chr.8: 143868828-143868828 on Build GRCh38
rs146005478 Chr.8: 143868744-143868744 on Build GRCh38
rs1054923745 Chr.8: 143868672-143868672 on Build GRCh38
rs782143817 Chr.8: 143868767-143868767 on Build GRCh38
rs782112444 Chr.8: 143868820-143868820 on Build GRCh38
rs551340158 Chr.8: 143868809-143868809 on Build GRCh38
rs782051022 Chr.8: 143868699-143868699 on Build GRCh38
rs782125557 Chr.8: 143868690-143868690 on Build GRCh38
rs573015971 Chr.8: 143868664-143868664 on Build GRCh38
rs782364262 Chr.8: 143868647-143868647 on Build GRCh38
rs782623089 Chr.8: 143868731-143868731 on Build GRCh38
rs35301416 Chr.8: 143868659-143868660 on Build GRCh38
rs782727181 Chr.8: 143868795-143868795 on Build GRCh38
rs782354838 Chr.8: 143868811-143868811 on Build GRCh38
rs782125308 Chr.8: 143868783-143868783 on Build GRCh38
rs782064600 Chr.8: 143868675-143868675 on Build GRCh38
rs782373090 Chr.8: 143868745-143868745 on Build GRCh38
rs782093131 Chr.8: 143868759-143868759 on Build GRCh38
rs201986967 Chr.8: 143868646-143868646 on Build GRCh38
rs894986070 Chr.8: 143868673-143868673 on Build GRCh38
rs373861393 Chr.8: 143868681-143868681 on Build GRCh38
rs782477999 Chr.8: 143868727-143868727 on Build GRCh38
rs781958862 Chr.8: 143868823-143868823 on Build GRCh38
rs267601818 Chr.8: 143868682-143868682 on Build GRCh38
rs964881215 Chr.8: 143868764-143868764 on Build GRCh38
rs782191264 Chr.8: 143868732-143868732 on Build GRCh38
rs114155737 Chr.8: 143868766-143868766 on Build GRCh38
rs367640883 Chr.8: 143868804-143868804 on Build GRCh38
rs781951311 Chr.8: 143868752-143868752 on Build GRCh38
rs529491804 Chr.8: 143868736-143868736 on Build GRCh38
rs781847075 Chr.8: 143868782-143868782 on Build GRCh38
rs373681462 Chr.8: 143868640-143868640 on Build GRCh38
rs377277154 Chr.8: 143868816-143868816 on Build GRCh38
rs201982226 Chr.8: 143868807-143868807 on Build GRCh38
rs370414140 Chr.8: 143868655-143868655 on Build GRCh38
rs190800981 Chr.8: 143868660-143868660 on Build GRCh38
rs782205400 Chr.8: 143868810-143868810 on Build GRCh38
rs372624077 Chr.8: 143868806-143868806 on Build GRCh38

View Full Product Details