Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151014003-151014276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG9B
Entrez Gene ID
285973
Gene Name
autophagy related 9B
Gene Aliases
APG9L2, NOS3AS, SONE
UniGene
Hs.707300
Gene Chromosome Location
Chr.7: 151012209-151024499 on Build GRCh38
Transcripts
NR_073169.1 NR_133652.1 XM_011516066.2 NM_000603.4 XM_017012232.1 XM_017012233.1

Gene Symbol
NOS3
Entrez Gene ID
4846
Gene Name
nitric oxide synthase 3
Gene Aliases
ECNOS, eNOS
UniGene
Hs.647092
Gene Chromosome Location
Chr.7: 150991056-151014599 on Build GRCh38
Transcripts
NR_073169.1 NR_133652.1 XM_011516066.2 NM_000603.4 XM_017012232.1 XM_017012233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3879324 c.3480C>T p.F1160F Chr.7: 151014037-151014037 on Build GRCh38
4587251 c.3593C>T p.S1198L Chr.7: 151014150-151014150 on Build GRCh38
5968337 c.3481G>T p.G1161W Chr.7: 151014038-151014038 on Build GRCh38
6331558 c.3552G>T p.Q1184H Chr.7: 151014109-151014109 on Build GRCh38
6331020 c.3507G>T p.V1169V Chr.7: 151014064-151014064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145823011 Chr.7: 151014104-151014104 on Build GRCh38
rs915666995 Chr.7: 151014136-151014136 on Build GRCh38
rs373133695 Chr.7: 151014145-151014145 on Build GRCh38
rs892696687 Chr.7: 151014141-151014141 on Build GRCh38
rs143054141 Chr.7: 151014055-151014055 on Build GRCh38
rs564937301 Chr.7: 151014140-151014140 on Build GRCh38
rs746325833 Chr.7: 151014150-151014150 on Build GRCh38
rs770226032 Chr.7: 151014113-151014113 on Build GRCh38
rs888441937 Chr.7: 151014032-151014032 on Build GRCh38
rs755842309 Chr.7: 151014082-151014082 on Build GRCh38
rs770846565 Chr.7: 151014147-151014147 on Build GRCh38
rs761225044 Chr.7: 151014118-151014118 on Build GRCh38
rs770455028 Chr.7: 151014156-151014156 on Build GRCh38
rs150714379 Chr.7: 151014031-151014031 on Build GRCh38
rs747320181 Chr.7: 151014146-151014146 on Build GRCh38
rs980965727 Chr.7: 151014178-151014178 on Build GRCh38
rs149818096 Chr.7: 151014101-151014101 on Build GRCh38
rs778093610 Chr.7: 151014093-151014093 on Build GRCh38
rs909247933 Chr.7: 151014224-151014224 on Build GRCh38
rs760123467 Chr.7: 151014108-151014108 on Build GRCh38
rs781344479 Chr.7: 151014148-151014148 on Build GRCh38
rs146141837 Chr.7: 151014071-151014071 on Build GRCh38
rs773809579 Chr.7: 151014181-151014181 on Build GRCh38
rs779787527 Chr.7: 151014123-151014123 on Build GRCh38
rs769133349 Chr.7: 151014177-151014177 on Build GRCh38
rs757520710 Chr.7: 151014215-151014215 on Build GRCh38
rs761543450 Chr.7: 151014192-151014192 on Build GRCh38
rs1022405149 Chr.7: 151014164-151014164 on Build GRCh38
rs750442776 Chr.7: 151014072-151014072 on Build GRCh38
rs772315573 Chr.7: 151014092-151014092 on Build GRCh38
rs748323632 Chr.7: 151014090-151014090 on Build GRCh38
rs151197128 Chr.7: 151014175-151014175 on Build GRCh38
rs753827185 Chr.7: 151014210-151014210 on Build GRCh38
rs368256841 Chr.7: 151014206-151014206 on Build GRCh38
rs1010583238 Chr.7: 151014157-151014157 on Build GRCh38
rs139120796 Chr.7: 151014040-151014040 on Build GRCh38
rs781380889 Chr.7: 151014218-151014218 on Build GRCh38
rs376955666 Chr.7: 151014198-151014198 on Build GRCh38
rs749398455 Chr.7: 151014117-151014117 on Build GRCh38
rs142886510 Chr.7: 151014119-151014119 on Build GRCh38
rs779840976 Chr.7: 151014088-151014088 on Build GRCh38
rs753603291 Chr.7: 151014124-151014124 on Build GRCh38
rs529527080 Chr.7: 151014251-151014251 on Build GRCh38
rs766891052 Chr.7: 151014194-151014194 on Build GRCh38
rs148530358 Chr.7: 151014115-151014115 on Build GRCh38
rs756842629 Chr.7: 151014066-151014066 on Build GRCh38
rs777840587 Chr.7: 151014143-151014143 on Build GRCh38
rs754722607 Chr.7: 151014211-151014211 on Build GRCh38
rs541336826 Chr.7: 151014116-151014116 on Build GRCh38
rs776189124 Chr.7: 151014174-151014174 on Build GRCh38
rs140184474 Chr.7: 151014095-151014095 on Build GRCh38
rs775291992 Chr.7: 151014162-151014162 on Build GRCh38
rs141211712 Chr.7: 151014025-151014025 on Build GRCh38
rs199762416 Chr.7: 151014213-151014213 on Build GRCh38

View Full Product Details