Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116858913-116859180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
NM_001242699.1 XM_005269815.3 XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6371278 c.581C>T p.P194L Chr.10: 116859082-116859082 on Build GRCh38
6368392 c.546C>A p.S182R Chr.10: 116859047-116859047 on Build GRCh38
2054867 c.499G>A p.A167T Chr.10: 116859000-116859000 on Build GRCh38
6843308 c.532G>C p.E178Q Chr.10: 116859033-116859033 on Build GRCh38
4972124 c.616delA p.K208fs*32 Chr.10: 116859117-116859117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187849003 Chr.10: 116859012-116859012 on Build GRCh38
rs759244496 Chr.10: 116859149-116859149 on Build GRCh38
rs912323689 Chr.10: 116859087-116859087 on Build GRCh38
rs368340847 Chr.10: 116859018-116859018 on Build GRCh38
rs936601022 Chr.10: 116859118-116859118 on Build GRCh38
rs941518429 Chr.10: 116859100-116859100 on Build GRCh38
rs769068676 Chr.10: 116859086-116859088 on Build GRCh38
rs150721071 Chr.10: 116859099-116859099 on Build GRCh38
rs982917268 Chr.10: 116859043-116859043 on Build GRCh38
rs975685135 Chr.10: 116859108-116859108 on Build GRCh38
rs921093412 Chr.10: 116859116-116859116 on Build GRCh38
rs760327781 Chr.10: 116859081-116859081 on Build GRCh38
rs370552301 Chr.10: 116858984-116858984 on Build GRCh38
rs995718247 Chr.10: 116858999-116858999 on Build GRCh38
rs199571664 Chr.10: 116859143-116859143 on Build GRCh38
rs368025714 Chr.10: 116859000-116859000 on Build GRCh38
rs771141549 Chr.10: 116858963-116858963 on Build GRCh38
rs774704085 Chr.10: 116859117-116859117 on Build GRCh38
rs761717176 Chr.10: 116859116-116859117 on Build GRCh38
rs765955713 Chr.10: 116859134-116859134 on Build GRCh38
rs989330355 Chr.10: 116859133-116859133 on Build GRCh38
rs957853121 Chr.10: 116858957-116858957 on Build GRCh38
rs759636926 Chr.10: 116859037-116859037 on Build GRCh38
rs776738848 Chr.10: 116858997-116858997 on Build GRCh38
rs989599721 Chr.10: 116858982-116858982 on Build GRCh38
rs542492628 Chr.10: 116859032-116859032 on Build GRCh38
rs914593863 Chr.10: 116859136-116859136 on Build GRCh38

View Full Product Details