Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13928105-13928376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2143618 c.692A>G p.D231G Chr.16: 13928135-13928135 on Build GRCh38
6271666 c.670G>T p.A224S Chr.16: 13928113-13928113 on Build GRCh38
1205614 c.770C>T p.A257V Chr.16: 13928213-13928213 on Build GRCh38
6522593 c.751G>A p.D251N Chr.16: 13928194-13928194 on Build GRCh38
967191 c.737C>T p.S246L Chr.16: 13928180-13928180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746904084 Chr.16: 13928161-13928161 on Build GRCh38
rs755368456 Chr.16: 13928145-13928145 on Build GRCh38
rs141101671 Chr.16: 13928146-13928146 on Build GRCh38
rs760444530 Chr.16: 13928197-13928197 on Build GRCh38
rs969667669 Chr.16: 13928131-13928131 on Build GRCh38
rs988493104 Chr.16: 13928331-13928331 on Build GRCh38
rs765111229 Chr.16: 13928225-13928225 on Build GRCh38
rs539438893 Chr.16: 13928301-13928301 on Build GRCh38
rs754532363 Chr.16: 13928277-13928277 on Build GRCh38
rs956186369 Chr.16: 13928316-13928316 on Build GRCh38
rs776221530 Chr.16: 13928199-13928199 on Build GRCh38
rs140299888 Chr.16: 13928219-13928219 on Build GRCh38
rs757874529 Chr.16: 13928257-13928257 on Build GRCh38
rs754843955 Chr.16: 13928242-13928243 on Build GRCh38
rs397509402 Chr.16: 13928132-13928132 on Build GRCh38
rs749985423 Chr.16: 13928245-13928245 on Build GRCh38
rs779737289 Chr.16: 13928129-13928129 on Build GRCh38
rs368218302 Chr.16: 13928143-13928143 on Build GRCh38
rs908295786 Chr.16: 13928186-13928186 on Build GRCh38
rs144608823 Chr.16: 13928171-13928171 on Build GRCh38
rs397509403 Chr.16: 13928149-13928149 on Build GRCh38
rs899417305 Chr.16: 13928142-13928142 on Build GRCh38
rs368789680 Chr.16: 13928258-13928258 on Build GRCh38
rs748488122 Chr.16: 13928286-13928286 on Build GRCh38
rs751290625 Chr.16: 13928226-13928226 on Build GRCh38
rs749352103 Chr.16: 13928179-13928179 on Build GRCh38
rs751214491 Chr.16: 13928266-13928266 on Build GRCh38
rs780166871 Chr.16: 13928157-13928157 on Build GRCh38
rs777855055 Chr.16: 13928248-13928252 on Build GRCh38
rs754173289 Chr.16: 13928249-13928249 on Build GRCh38
rs768217559 Chr.16: 13928168-13928168 on Build GRCh38
rs757357288 Chr.16: 13928270-13928270 on Build GRCh38
rs781626360 Chr.16: 13928285-13928285 on Build GRCh38
rs146650135 Chr.16: 13928181-13928181 on Build GRCh38
rs763961422 Chr.16: 13928198-13928198 on Build GRCh38
rs773444862 Chr.16: 13928176-13928176 on Build GRCh38
rs370864937 Chr.16: 13928180-13928180 on Build GRCh38
rs201176880 Chr.16: 13928173-13928173 on Build GRCh38
rs962536419 Chr.16: 13928212-13928212 on Build GRCh38

View Full Product Details