Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5733256-5733529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
UniGene
Hs.646899
Gene Chromosome Location
Chr.4: 5711197-5829043 on Build GRCh38
Transcripts
NM_001306090.1 NM_001306092.1 NM_153717.2 XM_006713865.3 XM_006713866.3 XM_011513419.2 XM_017007883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471703 c.660C>T p.D220D Chr.4: 5733393-5733393 on Build GRCh38
3604657 c.633G>A p.L211L Chr.4: 5733366-5733366 on Build GRCh38
1430358 c.679G>A p.A227T Chr.4: 5733412-5733412 on Build GRCh38
179279 c.625G>A p.V209I Chr.4: 5733358-5733358 on Build GRCh38
310952 c.641A>G p.Y214C Chr.4: 5733374-5733374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375549709 Chr.4: 5733288-5733288 on Build GRCh38
rs746235321 Chr.4: 5733384-5733384 on Build GRCh38
rs922198372 Chr.4: 5733430-5733430 on Build GRCh38
rs767274362 Chr.4: 5733337-5733337 on Build GRCh38
rs112148675 Chr.4: 5733476-5733476 on Build GRCh38
rs772621806 Chr.4: 5733411-5733411 on Build GRCh38
rs185642528 Chr.4: 5733459-5733459 on Build GRCh38
rs776878680 Chr.4: 5733314-5733314 on Build GRCh38
rs201796001 Chr.4: 5733406-5733406 on Build GRCh38
rs376818413 Chr.4: 5733361-5733361 on Build GRCh38
rs559719173 Chr.4: 5733452-5733452 on Build GRCh38
rs563481672 Chr.4: 5733423-5733423 on Build GRCh38
rs748795383 Chr.4: 5733489-5733489 on Build GRCh38
rs769320341 Chr.4: 5733461-5733461 on Build GRCh38
rs374437149 Chr.4: 5733310-5733310 on Build GRCh38
rs187457187 Chr.4: 5733357-5733357 on Build GRCh38
rs770326524 Chr.4: 5733453-5733453 on Build GRCh38
rs776989981 Chr.4: 5733383-5733383 on Build GRCh38
rs201583621 Chr.4: 5733410-5733410 on Build GRCh38
rs180960931 Chr.4: 5733451-5733451 on Build GRCh38
rs758352741 Chr.4: 5733433-5733433 on Build GRCh38
rs772174934 Chr.4: 5733370-5733370 on Build GRCh38
rs190661824 Chr.4: 5733435-5733435 on Build GRCh38
rs16837599 Chr.4: 5733462-5733462 on Build GRCh38
rs202026284 Chr.4: 5733358-5733358 on Build GRCh38
rs373002232 Chr.4: 5733379-5733379 on Build GRCh38
rs2291156 Chr.4: 5733329-5733329 on Build GRCh38
rs747203423 Chr.4: 5733378-5733378 on Build GRCh38
rs766045992 Chr.4: 5733481-5733481 on Build GRCh38
rs369085014 Chr.4: 5733457-5733457 on Build GRCh38
rs770414518 Chr.4: 5733394-5733394 on Build GRCh38
rs771487366 Chr.4: 5733309-5733309 on Build GRCh38
rs760051144 Chr.4: 5733323-5733323 on Build GRCh38
rs371664035 Chr.4: 5733429-5733429 on Build GRCh38
rs181951579 Chr.4: 5733280-5733280 on Build GRCh38
rs372274961 Chr.4: 5733333-5733333 on Build GRCh38
rs145945359 Chr.4: 5733330-5733330 on Build GRCh38
rs933295872 Chr.4: 5733440-5733440 on Build GRCh38
rs891834997 Chr.4: 5733470-5733470 on Build GRCh38
rs959703059 Chr.4: 5733298-5733298 on Build GRCh38
rs769431829 Chr.4: 5733431-5733431 on Build GRCh38
rs776205592 Chr.4: 5733401-5733401 on Build GRCh38
rs372090116 Chr.4: 5733308-5733308 on Build GRCh38
rs760158352 Chr.4: 5733473-5733473 on Build GRCh38
rs777946797 Chr.4: 5733373-5733373 on Build GRCh38
rs771372595 Chr.4: 5733311-5733311 on Build GRCh38
rs757444170 Chr.4: 5733443-5733443 on Build GRCh38
rs200294295 Chr.4: 5733393-5733393 on Build GRCh38
rs141979471 Chr.4: 5733285-5733285 on Build GRCh38
rs1043186520 Chr.4: 5733500-5733500 on Build GRCh38
rs982677323 Chr.4: 5733365-5733365 on Build GRCh38
rs778879455 Chr.4: 5733366-5733366 on Build GRCh38
rs765958700 Chr.4: 5733417-5733417 on Build GRCh38
rs754792099 Chr.4: 5733427-5733427 on Build GRCh38
rs879069336 Chr.4: 5733493-5733493 on Build GRCh38
rs921048524 Chr.4: 5733469-5733469 on Build GRCh38
rs750141808 Chr.4: 5733350-5733350 on Build GRCh38
rs760306552 Chr.4: 5733415-5733415 on Build GRCh38

View Full Product Details