Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99878438-99878665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML1
Entrez Gene ID
2009
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
ELP79, EMAP, EMAPL, HuEMAP
UniGene
Hs.12451
Gene Chromosome Location
Chr.14: 99737699-99942060 on Build GRCh38
Transcripts
NM_001008707.1 NM_004434.2 XM_005267397.1 XM_005267398.3 XM_005267399.4 XM_005267400.4 XM_011536540.2 XM_011536541.1 XM_011536542.2 XM_017021074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
697113 c.466G>A p.E156K Chr.14: 99878510-99878510 on Build GRCh38
365048 c.491G>T p.R164M Chr.14: 99878535-99878535 on Build GRCh38
4499727 c.548C>T p.S183F Chr.14: 99878592-99878592 on Build GRCh38
469656 c.566A>G p.N189S Chr.14: 99878610-99878610 on Build GRCh38
7177091 c.482G>A p.G161E Chr.14: 99878526-99878526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771530759 Chr.14: 99878602-99878602 on Build GRCh38
rs772519043 Chr.14: 99878494-99878494 on Build GRCh38
rs779370794 Chr.14: 99878583-99878583 on Build GRCh38
rs1017963990 Chr.14: 99878465-99878465 on Build GRCh38
rs375177816 Chr.14: 99878476-99878476 on Build GRCh38
rs750230794 Chr.14: 99878565-99878565 on Build GRCh38
rs915212352 Chr.14: 99878468-99878468 on Build GRCh38
rs749660797 Chr.14: 99878574-99878574 on Build GRCh38
rs776900171 Chr.14: 99878526-99878526 on Build GRCh38
rs966697309 Chr.14: 99878470-99878470 on Build GRCh38
rs766560700 Chr.14: 99878521-99878521 on Build GRCh38
rs201185419 Chr.14: 99878570-99878570 on Build GRCh38
rs773781960 Chr.14: 99878478-99878478 on Build GRCh38
rs772800582 Chr.14: 99878621-99878621 on Build GRCh38
rs752770872 Chr.14: 99878544-99878544 on Build GRCh38
rs761071014 Chr.14: 99878514-99878514 on Build GRCh38
rs765414771 Chr.14: 99878536-99878536 on Build GRCh38
rs886037935 Chr.14: 99878513-99878513 on Build GRCh38
rs868758424 Chr.14: 99878629-99878629 on Build GRCh38
rs762320146 Chr.14: 99878488-99878488 on Build GRCh38
rs748693883 Chr.14: 99878592-99878592 on Build GRCh38
rs144315305 Chr.14: 99878529-99878529 on Build GRCh38
rs776210995 Chr.14: 99878632-99878632 on Build GRCh38
rs779637372 Chr.14: 99878571-99878571 on Build GRCh38
rs11846839 Chr.14: 99878628-99878628 on Build GRCh38
rs567226366 Chr.14: 99878497-99878497 on Build GRCh38
rs928193159 Chr.14: 99878553-99878553 on Build GRCh38
rs117862534 Chr.14: 99878633-99878633 on Build GRCh38
rs773480441 Chr.14: 99878496-99878496 on Build GRCh38
rs370894571 Chr.14: 99878466-99878466 on Build GRCh38
rs200678486 Chr.14: 99878581-99878581 on Build GRCh38
rs767527533 Chr.14: 99878548-99878548 on Build GRCh38
rs188341390 Chr.14: 99878627-99878627 on Build GRCh38
rs757286397 Chr.14: 99878545-99878545 on Build GRCh38
rs755880656 Chr.14: 99878634-99878634 on Build GRCh38

View Full Product Details