Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161591900-161592173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR2C
Entrez Gene ID
9103
Gene Name
Fc fragment of IgG receptor IIc (gene/pseudogene)
Gene Aliases
CD32, CD32C, CDW32, FCG2, FCRIIC, IGFR2
UniGene
Hs.656750
Gene Chromosome Location
Chr.1: 161581339-161601220 on Build GRCh38
Transcripts
NM_201563.5 NR_047648.1

SNP Detail

SNP ID Chromosome Location
rs868078371 Chr.1: 161591997-161591997 on Build GRCh38
rs755873822 Chr.1: 161592144-161592144 on Build GRCh38
rs777594828 Chr.1: 161592145-161592145 on Build GRCh38
rs867161903 Chr.1: 161592052-161592052 on Build GRCh38
rs1771555 Chr.1: 161591944-161591944 on Build GRCh38
rs906162346 Chr.1: 161592004-161592004 on Build GRCh38
rs1033882158 Chr.1: 161592115-161592115 on Build GRCh38
rs755901699 Chr.1: 161591940-161591940 on Build GRCh38
rs1047449507 Chr.1: 161591974-161591974 on Build GRCh38
rs553853687 Chr.1: 161592075-161592075 on Build GRCh38
rs71519227 Chr.1: 161591925-161591925 on Build GRCh38
rs562568693 Chr.1: 161592154-161592154 on Build GRCh38
rs61801811 Chr.1: 161591939-161591939 on Build GRCh38
rs574563742 Chr.1: 161591977-161591977 on Build GRCh38
rs545887300 Chr.1: 161592102-161592102 on Build GRCh38
rs894875224 Chr.1: 161592124-161592124 on Build GRCh38
rs116188507 Chr.1: 161591926-161591926 on Build GRCh38
rs1003303985 Chr.1: 161592103-161592103 on Build GRCh38
rs537779509 Chr.1: 161591927-161591927 on Build GRCh38
rs61801812 Chr.1: 161592042-161592042 on Build GRCh38
rs576961959 Chr.1: 161592076-161592076 on Build GRCh38
rs540013124 Chr.1: 161591978-161591978 on Build GRCh38

View Full Product Details