Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8040026-8040255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
UniGene
Hs.120844
Gene Chromosome Location
Chr.12: 8032680-8055522 on Build GRCh38
Transcripts
NM_018416.2 XM_011520760.2 XM_011520761.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687399 c.320G>C p.G107A Chr.12: 8040152-8040152 on Build GRCh38
4841183 c.273G>T p.E91D Chr.12: 8040105-8040105 on Build GRCh38
4211677 c.203G>A p.R68Q Chr.12: 8040035-8040035 on Build GRCh38
5903012 c.302C>T p.P101L Chr.12: 8040134-8040134 on Build GRCh38
3465983 c.242C>T p.S81F Chr.12: 8040074-8040074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965387632 Chr.12: 8040098-8040098 on Build GRCh38
rs759823116 Chr.12: 8040100-8040100 on Build GRCh38
rs945571125 Chr.12: 8040227-8040227 on Build GRCh38
rs374823580 Chr.12: 8040191-8040191 on Build GRCh38
rs377340764 Chr.12: 8040087-8040087 on Build GRCh38
rs12580604 Chr.12: 8040201-8040201 on Build GRCh38
rs193084823 Chr.12: 8040054-8040054 on Build GRCh38
rs757005116 Chr.12: 8040150-8040150 on Build GRCh38
rs763956521 Chr.12: 8040122-8040122 on Build GRCh38
rs763258765 Chr.12: 8040060-8040060 on Build GRCh38
rs756932068 Chr.12: 8040214-8040215 on Build GRCh38
rs1019306444 Chr.12: 8040086-8040086 on Build GRCh38
rs746500310 Chr.12: 8040192-8040192 on Build GRCh38
rs781695258 Chr.12: 8040049-8040049 on Build GRCh38
rs952617945 Chr.12: 8040193-8040193 on Build GRCh38
rs749748296 Chr.12: 8040211-8040211 on Build GRCh38
rs767775080 Chr.12: 8040111-8040111 on Build GRCh38
rs771262165 Chr.12: 8040084-8040084 on Build GRCh38
rs1013267549 Chr.12: 8040170-8040170 on Build GRCh38
rs538093373 Chr.12: 8040175-8040175 on Build GRCh38
rs774855324 Chr.12: 8040214-8040214 on Build GRCh38
rs754472539 Chr.12: 8040195-8040195 on Build GRCh38
rs760748528 Chr.12: 8040113-8040113 on Build GRCh38
rs200593250 Chr.12: 8040172-8040172 on Build GRCh38
rs771259531 Chr.12: 8040213-8040213 on Build GRCh38
rs552576369 Chr.12: 8040188-8040190 on Build GRCh38
rs143696902 Chr.12: 8040135-8040135 on Build GRCh38
rs758039940 Chr.12: 8040180-8040180 on Build GRCh38
rs201952649 Chr.12: 8040112-8040112 on Build GRCh38

View Full Product Details