Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152303232-152303457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123178 c.11446G>A p.E3816K Chr.1: 152303440-152303440 on Build GRCh38
6264856 c.11564G>C p.R3855T Chr.1: 152303322-152303322 on Build GRCh38
6540863 c.11651C>A p.S3884Y Chr.1: 152303235-152303235 on Build GRCh38
7238853 c.11548T>G p.S3850A Chr.1: 152303338-152303338 on Build GRCh38
896079 c.11610G>T p.E3870D Chr.1: 152303276-152303276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745787051 Chr.1: 152303283-152303283 on Build GRCh38
rs146684366 Chr.1: 152303421-152303421 on Build GRCh38
rs774898663 Chr.1: 152303403-152303403 on Build GRCh38
rs147067035 Chr.1: 152303314-152303314 on Build GRCh38
rs866267917 Chr.1: 152303331-152303331 on Build GRCh38
rs201615654 Chr.1: 152303410-152303410 on Build GRCh38
rs373589596 Chr.1: 152303344-152303344 on Build GRCh38
rs775846399 Chr.1: 152303370-152303370 on Build GRCh38
rs144022904 Chr.1: 152303319-152303319 on Build GRCh38
rs752111537 Chr.1: 152303432-152303432 on Build GRCh38
rs760739986 Chr.1: 152303371-152303371 on Build GRCh38
rs146524643 Chr.1: 152303305-152303305 on Build GRCh38
rs750193070 Chr.1: 152303375-152303375 on Build GRCh38
rs758437501 Chr.1: 152303379-152303379 on Build GRCh38
rs201288925 Chr.1: 152303396-152303396 on Build GRCh38
rs370872906 Chr.1: 152303402-152303402 on Build GRCh38
rs759664162 Chr.1: 152303356-152303356 on Build GRCh38
rs751517977 Chr.1: 152303387-152303387 on Build GRCh38
rs200181463 Chr.1: 152303327-152303327 on Build GRCh38
rs764179418 Chr.1: 152303414-152303414 on Build GRCh38
rs78434011 Chr.1: 152303333-152303333 on Build GRCh38
rs780944348 Chr.1: 152303343-152303343 on Build GRCh38
rs138437732 Chr.1: 152303420-152303420 on Build GRCh38
rs767080575 Chr.1: 152303430-152303430 on Build GRCh38
rs747753756 Chr.1: 152303345-152303345 on Build GRCh38
rs746355769 Chr.1: 152303406-152303406 on Build GRCh38
rs773840408 Chr.1: 152303310-152303310 on Build GRCh38
rs201811891 Chr.1: 152303349-152303349 on Build GRCh38
rs143233744 Chr.1: 152303350-152303350 on Build GRCh38
rs142115347 Chr.1: 152303264-152303264 on Build GRCh38
rs140349291 Chr.1: 152303270-152303270 on Build GRCh38
rs955580281 Chr.1: 152303366-152303366 on Build GRCh38
rs765093705 Chr.1: 152303372-152303372 on Build GRCh38
rs754412084 Chr.1: 152303339-152303339 on Build GRCh38
rs758352997 Chr.1: 152303329-152303329 on Build GRCh38
rs140464988 Chr.1: 152303407-152303407 on Build GRCh38
rs375448938 Chr.1: 152303322-152303322 on Build GRCh38
rs754572074 Chr.1: 152303388-152303388 on Build GRCh38
rs553373171 Chr.1: 152303299-152303299 on Build GRCh38
rs564072147 Chr.1: 152303351-152303351 on Build GRCh38
rs766454727 Chr.1: 152303383-152303383 on Build GRCh38
rs779638257 Chr.1: 152303288-152303288 on Build GRCh38
rs145079750 Chr.1: 152303400-152303400 on Build GRCh38
rs762274526 Chr.1: 152303304-152303304 on Build GRCh38
rs150496930 Chr.1: 152303334-152303334 on Build GRCh38
rs755138083 Chr.1: 152303253-152303253 on Build GRCh38
rs746671388 Chr.1: 152303291-152303291 on Build GRCh38
rs7537147 Chr.1: 152303401-152303401 on Build GRCh38
rs139279364 Chr.1: 152303392-152303392 on Build GRCh38
rs533030780 Chr.1: 152303398-152303398 on Build GRCh38
rs573388805 Chr.1: 152303311-152303311 on Build GRCh38
rs1057518212 Chr.1: 152303358-152303358 on Build GRCh38
rs199565469 Chr.1: 152303422-152303422 on Build GRCh38
rs778960079 Chr.1: 152303278-152303278 on Build GRCh38
rs751786975 Chr.1: 152303252-152303252 on Build GRCh38
rs776888744 Chr.1: 152303300-152303300 on Build GRCh38
rs930504039 Chr.1: 152303348-152303348 on Build GRCh38
rs867113991 Chr.1: 152303332-152303332 on Build GRCh38
rs777452476 Chr.1: 152303256-152303256 on Build GRCh38
rs149624484 Chr.1: 152303423-152303423 on Build GRCh38
rs74129447 Chr.1: 152303313-152303313 on Build GRCh38
rs750391074 Chr.1: 152303328-152303328 on Build GRCh38

View Full Product Details