Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155253753-155254024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189B
Entrez Gene ID
10712
Gene Name
family with sequence similarity 189 member B
Gene Aliases
C1orf2, COTE1
UniGene
Hs.348308
Gene Chromosome Location
Chr.1: 155247205-155255483 on Build GRCh38
Transcripts
NM_001267608.1 NM_006589.2 NM_198264.1 XM_005244845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1165459 c.452C>A p.S151* Chr.1: 155253920-155253920 on Build GRCh38
4807675 c.394-1G>C p.? Chr.1: 155253979-155253979 on Build GRCh38
3774409 c.513+10C>T p.? Chr.1: 155253849-155253849 on Build GRCh38
4022972 c.431T>C p.L144P Chr.1: 155253941-155253941 on Build GRCh38
4022971 c.468A>C p.K156N Chr.1: 155253904-155253904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750760284 Chr.1: 155253860-155253860 on Build GRCh38
rs776813703 Chr.1: 155253938-155253938 on Build GRCh38
rs757036171 Chr.1: 155253994-155253994 on Build GRCh38
rs373398871 Chr.1: 155253903-155253903 on Build GRCh38
rs758711310 Chr.1: 155253867-155253867 on Build GRCh38
rs777457335 Chr.1: 155253784-155253784 on Build GRCh38
rs1022315631 Chr.1: 155253872-155253872 on Build GRCh38
rs151056816 Chr.1: 155253850-155253850 on Build GRCh38
rs376213640 Chr.1: 155253939-155253939 on Build GRCh38
rs145460071 Chr.1: 155253920-155253920 on Build GRCh38
rs764229026 Chr.1: 155253830-155253830 on Build GRCh38
rs774269913 Chr.1: 155253922-155253922 on Build GRCh38
rs766916376 Chr.1: 155253958-155253958 on Build GRCh38
rs555235656 Chr.1: 155253787-155253787 on Build GRCh38
rs768701519 Chr.1: 155253935-155253935 on Build GRCh38
rs763340243 Chr.1: 155253953-155253953 on Build GRCh38
rs766934808 Chr.1: 155253875-155253875 on Build GRCh38
rs201759741 Chr.1: 155253963-155253963 on Build GRCh38
rs767100508 Chr.1: 155253967-155253967 on Build GRCh38
rs969959316 Chr.1: 155253970-155253970 on Build GRCh38
rs2990232 Chr.1: 155253799-155253799 on Build GRCh38
rs761554594 Chr.1: 155253825-155253825 on Build GRCh38
rs558216744 Chr.1: 155253856-155253856 on Build GRCh38
rs747731053 Chr.1: 155253907-155253907 on Build GRCh38
rs538015677 Chr.1: 155253927-155253927 on Build GRCh38
rs770811101 Chr.1: 155253814-155253814 on Build GRCh38
rs773319049 Chr.1: 155253945-155253945 on Build GRCh38
rs777421608 Chr.1: 155253981-155253981 on Build GRCh38
rs752114822 Chr.1: 155253882-155253882 on Build GRCh38
rs774290111 Chr.1: 155253818-155253818 on Build GRCh38
rs369560740 Chr.1: 155253889-155253889 on Build GRCh38
rs200280419 Chr.1: 155253971-155253971 on Build GRCh38
rs773090959 Chr.1: 155253999-155253999 on Build GRCh38
rs755826787 Chr.1: 155253918-155253918 on Build GRCh38
rs752096458 Chr.1: 155253961-155253961 on Build GRCh38
rs1022800890 Chr.1: 155253960-155253960 on Build GRCh38
rs971257763 Chr.1: 155253821-155253821 on Build GRCh38
rs760684304 Chr.1: 155253822-155253822 on Build GRCh38
rs749057195 Chr.1: 155253919-155253919 on Build GRCh38
rs745867271 Chr.1: 155253925-155253925 on Build GRCh38
rs753994823 Chr.1: 155253847-155253847 on Build GRCh38
rs201271382 Chr.1: 155253783-155253783 on Build GRCh38
rs748962225 Chr.1: 155253987-155253987 on Build GRCh38
rs773104190 Chr.1: 155253780-155253780 on Build GRCh38

View Full Product Details