Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27614375-27614618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGR
Entrez Gene ID
2268
Gene Name
FGR proto-oncogene, Src family tyrosine kinase
Gene Aliases
SRC2, c-fgr, c-src2, p55-Fgr, p55c-fgr, p58-Fgr, p58c-fgr
UniGene
Hs.1422
Gene Chromosome Location
Chr.1: 27612289-27635561 on Build GRCh38
Transcripts
NM_001042729.1 NM_001042747.1 NM_005248.2 XM_006710452.2 XM_011541010.1 XM_011541011.2 XM_011541012.1 XM_011541013.2 XM_011541014.2 XM_017000673.1 XM_017000674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907904 c.1130A>G p.N377S Chr.1: 27614549-27614549 on Build GRCh38
3360812 c.1098A>G p.V366V Chr.1: 27614581-27614581 on Build GRCh38
7278530 c.1124G>A p.R375H Chr.1: 27614555-27614555 on Build GRCh38
4408278 c.1141C>T p.R381C Chr.1: 27614538-27614538 on Build GRCh38
1627001 c.1152G>A p.R384R Chr.1: 27614527-27614527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936244720 Chr.1: 27614494-27614494 on Build GRCh38
rs779390995 Chr.1: 27614501-27614501 on Build GRCh38
rs879373811 Chr.1: 27614555-27614555 on Build GRCh38
rs567390006 Chr.1: 27614400-27614400 on Build GRCh38
rs755738879 Chr.1: 27614468-27614468 on Build GRCh38
rs775661836 Chr.1: 27614425-27614425 on Build GRCh38
rs772494756 Chr.1: 27614416-27614416 on Build GRCh38
rs143532619 Chr.1: 27614524-27614524 on Build GRCh38
rs780417214 Chr.1: 27614503-27614503 on Build GRCh38
rs141214383 Chr.1: 27614495-27614495 on Build GRCh38
rs776733746 Chr.1: 27614438-27614438 on Build GRCh38
rs752268862 Chr.1: 27614465-27614465 on Build GRCh38
rs767580735 Chr.1: 27614564-27614564 on Build GRCh38
rs767460030 Chr.1: 27614458-27614458 on Build GRCh38
rs56377124 Chr.1: 27614536-27614536 on Build GRCh38
rs138858537 Chr.1: 27614508-27614508 on Build GRCh38
rs777318062 Chr.1: 27614493-27614493 on Build GRCh38
rs757288230 Chr.1: 27614586-27614586 on Build GRCh38
rs372083839 Chr.1: 27614538-27614538 on Build GRCh38
rs748767733 Chr.1: 27614397-27614397 on Build GRCh38
rs372569371 Chr.1: 27614415-27614415 on Build GRCh38
rs761805327 Chr.1: 27614439-27614439 on Build GRCh38
rs766362704 Chr.1: 27614443-27614443 on Build GRCh38
rs770064698 Chr.1: 27614546-27614546 on Build GRCh38
rs1041850783 Chr.1: 27614434-27614434 on Build GRCh38
rs759399215 Chr.1: 27614456-27614456 on Build GRCh38
rs907261374 Chr.1: 27614574-27614574 on Build GRCh38
rs746418519 Chr.1: 27614502-27614502 on Build GRCh38
rs924897313 Chr.1: 27614452-27614452 on Build GRCh38
rs774841609 Chr.1: 27614404-27614404 on Build GRCh38
rs768782391 Chr.1: 27614437-27614437 on Build GRCh38
rs751602520 Chr.1: 27614446-27614446 on Build GRCh38
rs761010060 Chr.1: 27614427-27614427 on Build GRCh38
rs774399386 Chr.1: 27614557-27614557 on Build GRCh38
rs752493866 Chr.1: 27614583-27614583 on Build GRCh38
rs753758593 Chr.1: 27614544-27614545 on Build GRCh38
rs756507665 Chr.1: 27614537-27614537 on Build GRCh38
rs746220634 Chr.1: 27614411-27614411 on Build GRCh38
rs760387692 Chr.1: 27614593-27614593 on Build GRCh38
rs759595392 Chr.1: 27614562-27614562 on Build GRCh38
rs548710886 Chr.1: 27614515-27614515 on Build GRCh38

View Full Product Details