Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74800693-74800942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM169A
Entrez Gene ID
26049
Gene Name
family with sequence similarity 169 member A
Gene Aliases
SLAP75
UniGene
Hs.91662
Gene Chromosome Location
Chr.5: 74777574-74867994 on Build GRCh38
Transcripts
NM_015566.2 NR_046462.1 XM_005248480.2 XM_011543306.2 XM_011543307.2 XM_017009332.1 XM_017009333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5047359 c.1088C>T p.T363I Chr.5: 74800895-74800895 on Build GRCh38
5502975 c.1043G>A p.S348N Chr.5: 74800940-74800940 on Build GRCh38
6836514 c.1043G>C p.S348T Chr.5: 74800940-74800940 on Build GRCh38
5615563 c.1098A>G p.I366M Chr.5: 74800885-74800885 on Build GRCh38
5403806 c.1063G>A p.E355K Chr.5: 74800920-74800920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761414506 Chr.5: 74800837-74800837 on Build GRCh38
rs768325367 Chr.5: 74800848-74800848 on Build GRCh38
rs766795583 Chr.5: 74800891-74800891 on Build GRCh38
rs374275749 Chr.5: 74800914-74800914 on Build GRCh38
rs755287315 Chr.5: 74800902-74800902 on Build GRCh38
rs950869770 Chr.5: 74800808-74800810 on Build GRCh38
rs374078379 Chr.5: 74800842-74800842 on Build GRCh38
rs200900016 Chr.5: 74800875-74800875 on Build GRCh38
rs776299584 Chr.5: 74800860-74800861 on Build GRCh38
rs765765816 Chr.5: 74800904-74800904 on Build GRCh38
rs984953086 Chr.5: 74800810-74800810 on Build GRCh38
rs894220342 Chr.5: 74800895-74800895 on Build GRCh38
rs1038658968 Chr.5: 74800909-74800909 on Build GRCh38
rs537814895 Chr.5: 74800776-74800776 on Build GRCh38
rs766905048 Chr.5: 74800841-74800841 on Build GRCh38
rs992813299 Chr.5: 74800720-74800720 on Build GRCh38
rs919551148 Chr.5: 74800751-74800751 on Build GRCh38
rs760389519 Chr.5: 74800761-74800764 on Build GRCh38
rs372048094 Chr.5: 74800912-74800912 on Build GRCh38
rs34252559 Chr.5: 74800766-74800766 on Build GRCh38
rs908937741 Chr.5: 74800811-74800811 on Build GRCh38

View Full Product Details