Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57551226-57551487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FECH
Entrez Gene ID
2235
Gene Name
ferrochelatase
Gene Aliases
EPP, FCE
UniGene
Hs.365365
Gene Chromosome Location
Chr.18: 57544841-57586737 on Build GRCh38
Transcripts
NM_000140.3 NM_001012515.2 XM_011525881.1 XM_011525882.2 XM_017025614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
989132 c.1142C>A p.P381Q Chr.18: 57551328-57551328 on Build GRCh38
141528 c.1130T>C p.L377P Chr.18: 57551340-57551340 on Build GRCh38
5531197 c.1129C>T p.L377F Chr.18: 57551341-57551341 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779317985 Chr.18: 57551282-57551282 on Build GRCh38
rs888114623 Chr.18: 57551356-57551356 on Build GRCh38
rs764466739 Chr.18: 57551316-57551316 on Build GRCh38
rs755462599 Chr.18: 57551276-57551276 on Build GRCh38
rs774797955 Chr.18: 57551412-57551416 on Build GRCh38
rs965601057 Chr.18: 57551433-57551433 on Build GRCh38
rs879255507 Chr.18: 57551317-57551317 on Build GRCh38
rs202147607 Chr.18: 57551312-57551312 on Build GRCh38
rs186440873 Chr.18: 57551294-57551294 on Build GRCh38
rs369940306 Chr.18: 57551267-57551267 on Build GRCh38
rs377190928 Chr.18: 57551297-57551297 on Build GRCh38
rs750958443 Chr.18: 57551424-57551425 on Build GRCh38
rs796643612 Chr.18: 57551433-57551433 on Build GRCh38
rs368714845 Chr.18: 57551310-57551310 on Build GRCh38
rs762793549 Chr.18: 57551369-57551369 on Build GRCh38
rs896295055 Chr.18: 57551393-57551393 on Build GRCh38
rs202130606 Chr.18: 57551336-57551336 on Build GRCh38
rs867651405 Chr.18: 57551341-57551341 on Build GRCh38
rs763237544 Chr.18: 57551307-57551307 on Build GRCh38
rs746593629 Chr.18: 57551299-57551299 on Build GRCh38
rs763940453 Chr.18: 57551385-57551385 on Build GRCh38
rs780037119 Chr.18: 57551263-57551263 on Build GRCh38
rs57800921 Chr.18: 57551425-57551425 on Build GRCh38
rs753913557 Chr.18: 57551425-57551425 on Build GRCh38
rs1048121841 Chr.18: 57551327-57551327 on Build GRCh38
rs373537776 Chr.18: 57551413-57551413 on Build GRCh38
rs762241893 Chr.18: 57551419-57551419 on Build GRCh38
rs118204040 Chr.18: 57551367-57551367 on Build GRCh38
rs772379635 Chr.18: 57551295-57551295 on Build GRCh38
rs770720963 Chr.18: 57551308-57551308 on Build GRCh38
rs980162834 Chr.18: 57551278-57551278 on Build GRCh38
rs554319418 Chr.18: 57551448-57551448 on Build GRCh38
rs751356941 Chr.18: 57551408-57551408 on Build GRCh38
rs201722847 Chr.18: 57551419-57551419 on Build GRCh38
rs556144432 Chr.18: 57551319-57551319 on Build GRCh38

View Full Product Details