Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3977926-3978175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2
Entrez Gene ID
1938
Gene Name
eukaryotic translation elongation factor 2
Gene Aliases
EEF-2, EF-2, EF2, SCA26
UniGene
Hs.515070
Gene Chromosome Location
Chr.19: 3976056-3985463 on Build GRCh38
Transcripts
NM_001961.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5024848 c.1724C>A p.P575Q Chr.19: 3978162-3978162 on Build GRCh38
122778 c.1898G>A p.R633H Chr.19: 3977988-3977988 on Build GRCh38
4425853 c.1796A>G p.H599R Chr.19: 3978090-3978090 on Build GRCh38
1612217 c.1905G>T p.L635L Chr.19: 3977981-3977981 on Build GRCh38
6651763 c.1896G>A p.A632A Chr.19: 3977990-3977990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746161915 Chr.19: 3977975-3977975 on Build GRCh38
rs754786270 Chr.19: 3977977-3977977 on Build GRCh38
rs971710297 Chr.19: 3978152-3978152 on Build GRCh38
rs200515602 Chr.19: 3977969-3977969 on Build GRCh38
rs1007902036 Chr.19: 3978055-3978055 on Build GRCh38
rs777751138 Chr.19: 3978035-3978035 on Build GRCh38
rs768970441 Chr.19: 3978076-3978076 on Build GRCh38
rs755946485 Chr.19: 3978034-3978034 on Build GRCh38
rs587777052 Chr.19: 3978099-3978099 on Build GRCh38
rs1029194030 Chr.19: 3977951-3977951 on Build GRCh38
rs755931480 Chr.19: 3978129-3978129 on Build GRCh38
rs753183188 Chr.19: 3977959-3977959 on Build GRCh38
rs767911696 Chr.19: 3977948-3977948 on Build GRCh38
rs779267368 Chr.19: 3977973-3977973 on Build GRCh38
rs373868450 Chr.19: 3978149-3978149 on Build GRCh38
rs370836485 Chr.19: 3978125-3978125 on Build GRCh38
rs966389526 Chr.19: 3978079-3978079 on Build GRCh38
rs148878859 Chr.19: 3978140-3978140 on Build GRCh38
rs772529820 Chr.19: 3977983-3977983 on Build GRCh38
rs773988050 Chr.19: 3977991-3977991 on Build GRCh38
rs759879199 Chr.19: 3977945-3977945 on Build GRCh38
rs147383237 Chr.19: 3977960-3977960 on Build GRCh38
rs139427550 Chr.19: 3977978-3977978 on Build GRCh38
rs754364920 Chr.19: 3978013-3978013 on Build GRCh38
rs781090546 Chr.19: 3978026-3978026 on Build GRCh38
rs377744684 Chr.19: 3978043-3978043 on Build GRCh38
rs997431543 Chr.19: 3978036-3978036 on Build GRCh38
rs777167717 Chr.19: 3978101-3978101 on Build GRCh38
rs201326053 Chr.19: 3978050-3978050 on Build GRCh38
rs757356247 Chr.19: 3978016-3978016 on Build GRCh38
rs35987350 Chr.19: 3978121-3978121 on Build GRCh38
rs770242626 Chr.19: 3978044-3978044 on Build GRCh38
rs145009005 Chr.19: 3978005-3978005 on Build GRCh38
rs368234090 Chr.19: 3978068-3978068 on Build GRCh38
rs145540039 Chr.19: 3978116-3978116 on Build GRCh38
rs139615561 Chr.19: 3978056-3978056 on Build GRCh38
rs764042018 Chr.19: 3977968-3977968 on Build GRCh38
rs980995029 Chr.19: 3978010-3978010 on Build GRCh38
rs149757304 Chr.19: 3978066-3978066 on Build GRCh38
rs200589086 Chr.19: 3977955-3977955 on Build GRCh38
rs773875633 Chr.19: 3978051-3978051 on Build GRCh38
rs867219501 Chr.19: 3977946-3977946 on Build GRCh38
rs373062093 Chr.19: 3977990-3977990 on Build GRCh38
rs201976284 Chr.19: 3977994-3977994 on Build GRCh38
rs775864126 Chr.19: 3978002-3978002 on Build GRCh38
rs147570651 Chr.19: 3978017-3978017 on Build GRCh38
rs750648849 Chr.19: 3978018-3978018 on Build GRCh38
rs979060977 Chr.19: 3978095-3978095 on Build GRCh38
rs375212891 Chr.19: 3978053-3978053 on Build GRCh38
rs1016486852 Chr.19: 3978148-3978148 on Build GRCh38
rs758714357 Chr.19: 3978020-3978020 on Build GRCh38
rs200869338 Chr.19: 3978107-3978107 on Build GRCh38
rs149189348 Chr.19: 3977981-3977981 on Build GRCh38
rs759237233 Chr.19: 3977993-3977993 on Build GRCh38
rs763197514 Chr.19: 3978122-3978122 on Build GRCh38
rs775329938 Chr.19: 3977972-3977972 on Build GRCh38
rs1020646538 Chr.19: 3978059-3978059 on Build GRCh38
rs1053673842 Chr.19: 3978049-3978049 on Build GRCh38

View Full Product Details