Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74879395-74879660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS6
Entrez Gene ID
283985
Gene Name
fatty acid desaturase 6
Gene Aliases
FP18279
UniGene
Hs.448845
Gene Chromosome Location
Chr.17: 74874793-74893811 on Build GRCh38
Transcripts
NM_178128.5 XM_005257224.4 XM_017024458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5530761 c.885C>T p.L295L Chr.17: 74879425-74879425 on Build GRCh38
1251686 c.843G>A p.S281S Chr.17: 74879467-74879467 on Build GRCh38
6147977 c.732C>T p.I244I Chr.17: 74879578-74879578 on Build GRCh38
2978139 c.766C>T p.R256C Chr.17: 74879544-74879544 on Build GRCh38
4682293 c.769C>T p.R257W Chr.17: 74879541-74879541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762778888 Chr.17: 74879493-74879493 on Build GRCh38
rs780492943 Chr.17: 74879559-74879559 on Build GRCh38
rs762441394 Chr.17: 74879530-74879530 on Build GRCh38
rs746966565 Chr.17: 74879482-74879482 on Build GRCh38
rs779079265 Chr.17: 74879544-74879544 on Build GRCh38
rs12945561 Chr.17: 74879620-74879620 on Build GRCh38
rs12952826 Chr.17: 74879534-74879534 on Build GRCh38
rs531632048 Chr.17: 74879480-74879480 on Build GRCh38
rs752697285 Chr.17: 74879608-74879608 on Build GRCh38
rs374369095 Chr.17: 74879479-74879479 on Build GRCh38
rs555267509 Chr.17: 74879580-74879580 on Build GRCh38
rs753087585 Chr.17: 74879605-74879605 on Build GRCh38
rs772664436 Chr.17: 74879586-74879586 on Build GRCh38
rs754039124 Chr.17: 74879529-74879529 on Build GRCh38
rs747238459 Chr.17: 74879631-74879631 on Build GRCh38
rs535361532 Chr.17: 74879578-74879578 on Build GRCh38
rs750485460 Chr.17: 74879616-74879616 on Build GRCh38
rs766378328 Chr.17: 74879527-74879527 on Build GRCh38
rs780182681 Chr.17: 74879630-74879630 on Build GRCh38
rs756928216 Chr.17: 74879476-74879476 on Build GRCh38
rs372130393 Chr.17: 74879494-74879494 on Build GRCh38
rs368208629 Chr.17: 74879481-74879481 on Build GRCh38
rs905353709 Chr.17: 74879437-74879437 on Build GRCh38
rs376084957 Chr.17: 74879612-74879612 on Build GRCh38
rs756050871 Chr.17: 74879609-74879609 on Build GRCh38
rs371433937 Chr.17: 74879475-74879475 on Build GRCh38
rs199975806 Chr.17: 74879540-74879540 on Build GRCh38
rs558921266 Chr.17: 74879563-74879563 on Build GRCh38
rs571618147 Chr.17: 74879626-74879626 on Build GRCh38
rs768913106 Chr.17: 74879632-74879632 on Build GRCh38
rs185169950 Chr.17: 74879526-74879526 on Build GRCh38
rs771101271 Chr.17: 74879496-74879496 on Build GRCh38
rs761030697 Chr.17: 74879441-74879441 on Build GRCh38
rs775654616 Chr.17: 74879503-74879503 on Build GRCh38
rs1002415494 Chr.17: 74879452-74879452 on Build GRCh38
rs758485509 Chr.17: 74879618-74879618 on Build GRCh38
rs367720047 Chr.17: 74879539-74879539 on Build GRCh38
rs769037014 Chr.17: 74879566-74879566 on Build GRCh38
rs1022968350 Chr.17: 74879587-74879587 on Build GRCh38
rs772500069 Chr.17: 74879502-74879502 on Build GRCh38
rs762689461 Chr.17: 74879596-74879596 on Build GRCh38
rs776122430 Chr.17: 74879431-74879431 on Build GRCh38
rs772148800 Chr.17: 74879558-74879558 on Build GRCh38
rs754830975 Chr.17: 74879489-74879489 on Build GRCh38
rs544027756 Chr.17: 74879599-74879599 on Build GRCh38
rs116982675 Chr.17: 74879597-74879597 on Build GRCh38
rs903514214 Chr.17: 74879615-74879615 on Build GRCh38
rs111943174 Chr.17: 74879554-74879554 on Build GRCh38
rs369986802 Chr.17: 74879543-74879543 on Build GRCh38
rs746075421 Chr.17: 74879501-74879501 on Build GRCh38
rs372859721 Chr.17: 74879606-74879606 on Build GRCh38
rs754155839 Chr.17: 74879530-74879531 on Build GRCh38
rs200986366 Chr.17: 74879517-74879517 on Build GRCh38
rs12945325 Chr.17: 74879589-74879589 on Build GRCh38
rs757535571 Chr.17: 74879559-74879560 on Build GRCh38
rs764226470 Chr.17: 74879485-74879487 on Build GRCh38
rs907161807 Chr.17: 74879588-74879588 on Build GRCh38
rs764461906 Chr.17: 74879521-74879521 on Build GRCh38
rs375603236 Chr.17: 74879577-74879577 on Build GRCh38
rs777102887 Chr.17: 74879524-74879524 on Build GRCh38
rs200416598 Chr.17: 74879509-74879509 on Build GRCh38
rs763669924 Chr.17: 74879468-74879468 on Build GRCh38
rs375851737 Chr.17: 74879541-74879541 on Build GRCh38

View Full Product Details