Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39401338-39401575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO33
Entrez Gene ID
254170
Gene Name
F-box protein 33
Gene Aliases
BMND12, Fbx33, c14_5247
UniGene
Hs.146632
Gene Chromosome Location
Chr.14: 39396373-39432500 on Build GRCh38
Transcripts
NM_203301.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3968811 c.1137C>T p.I379I Chr.14: 39401435-39401435 on Build GRCh38
6054437 c.1174A>G p.I392V Chr.14: 39401398-39401398 on Build GRCh38
7222104 c.1072G>C p.E358Q Chr.14: 39401500-39401500 on Build GRCh38
5612588 c.1170C>T p.P390P Chr.14: 39401402-39401402 on Build GRCh38
3496019 c.1051C>T p.L351L Chr.14: 39401521-39401521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771092232 Chr.14: 39401485-39401485 on Build GRCh38
rs1025466805 Chr.14: 39401543-39401543 on Build GRCh38
rs746414224 Chr.14: 39401511-39401511 on Build GRCh38
rs61754302 Chr.14: 39401390-39401390 on Build GRCh38
rs757152219 Chr.14: 39401363-39401363 on Build GRCh38
rs778980477 Chr.14: 39401365-39401365 on Build GRCh38
rs778035293 Chr.14: 39401478-39401478 on Build GRCh38
rs779846906 Chr.14: 39401461-39401461 on Build GRCh38
rs748313600 Chr.14: 39401471-39401471 on Build GRCh38
rs375157529 Chr.14: 39401552-39401552 on Build GRCh38
rs539795896 Chr.14: 39401526-39401526 on Build GRCh38
rs61754303 Chr.14: 39401528-39401528 on Build GRCh38
rs781363717 Chr.14: 39401371-39401371 on Build GRCh38
rs144987416 Chr.14: 39401435-39401435 on Build GRCh38
rs144152967 Chr.14: 39401417-39401417 on Build GRCh38
rs557650803 Chr.14: 39401421-39401421 on Build GRCh38
rs758089884 Chr.14: 39401460-39401460 on Build GRCh38
rs775239655 Chr.14: 39401434-39401434 on Build GRCh38
rs753286646 Chr.14: 39401439-39401439 on Build GRCh38
rs756852751 Chr.14: 39401447-39401447 on Build GRCh38
rs571077291 Chr.14: 39401430-39401430 on Build GRCh38
rs372862832 Chr.14: 39401398-39401398 on Build GRCh38
rs750418473 Chr.14: 39401454-39401454 on Build GRCh38
rs779256618 Chr.14: 39401495-39401495 on Build GRCh38
rs745641139 Chr.14: 39401367-39401367 on Build GRCh38
rs756335602 Chr.14: 39401472-39401472 on Build GRCh38
rs749313472 Chr.14: 39401483-39401483 on Build GRCh38
rs370540187 Chr.14: 39401544-39401544 on Build GRCh38
rs149090376 Chr.14: 39401529-39401529 on Build GRCh38
rs749738165 Chr.14: 39401410-39401410 on Build GRCh38
rs773294620 Chr.14: 39401399-39401399 on Build GRCh38
rs765068524 Chr.14: 39401452-39401452 on Build GRCh38
rs368046543 Chr.14: 39401393-39401393 on Build GRCh38
rs772511993 Chr.14: 39401514-39401514 on Build GRCh38

View Full Product Details