Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11069520-11069763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC10
Entrez Gene ID
5394
Gene Name
exosome component 10
Gene Aliases
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4
UniGene
Hs.632368
Gene Chromosome Location
Chr.1: 11066613-11099910 on Build GRCh38
Transcripts
NM_001001998.2 NM_002685.3 XM_005263475.2 XM_017001494.1 XM_017001495.1 XM_017001496.1 XM_017001497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6834994 c.2374C>T p.Q792* Chr.1: 11069673-11069673 on Build GRCh38
5034184 c.2475C>T p.F825F Chr.1: 11069572-11069572 on Build GRCh38
6657276 c.2393G>A p.R798Q Chr.1: 11069654-11069654 on Build GRCh38
3471107 c.2397C>T p.L799L Chr.1: 11069650-11069650 on Build GRCh38
302161 c.2449C>T p.P817S Chr.1: 11069598-11069598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200633625 Chr.1: 11069546-11069546 on Build GRCh38
rs150363329 Chr.1: 11069592-11069592 on Build GRCh38
rs184500057 Chr.1: 11069625-11069625 on Build GRCh38
rs751132534 Chr.1: 11069642-11069642 on Build GRCh38
rs755356177 Chr.1: 11069692-11069692 on Build GRCh38
rs765409707 Chr.1: 11069658-11069662 on Build GRCh38
rs149082475 Chr.1: 11069593-11069593 on Build GRCh38
rs779315339 Chr.1: 11069697-11069697 on Build GRCh38
rs775788127 Chr.1: 11069709-11069711 on Build GRCh38
rs773225231 Chr.1: 11069705-11069705 on Build GRCh38
rs749557592 Chr.1: 11069686-11069686 on Build GRCh38
rs61773684 Chr.1: 11069741-11069741 on Build GRCh38
rs758478602 Chr.1: 11069631-11069631 on Build GRCh38
rs759738982 Chr.1: 11069736-11069736 on Build GRCh38
rs897837626 Chr.1: 11069629-11069634 on Build GRCh38
rs776245730 Chr.1: 11069605-11069605 on Build GRCh38
rs143402418 Chr.1: 11069599-11069599 on Build GRCh38
rs773319678 Chr.1: 11069598-11069598 on Build GRCh38
rs534464112 Chr.1: 11069716-11069716 on Build GRCh38
rs369984967 Chr.1: 11069673-11069673 on Build GRCh38
rs780930127 Chr.1: 11069685-11069685 on Build GRCh38
rs952090050 Chr.1: 11069691-11069691 on Build GRCh38
rs776928606 Chr.1: 11069733-11069733 on Build GRCh38
rs759376992 Chr.1: 11069615-11069615 on Build GRCh38
rs766643284 Chr.1: 11069600-11069600 on Build GRCh38
rs780261334 Chr.1: 11069577-11069577 on Build GRCh38
rs549838469 Chr.1: 11069567-11069567 on Build GRCh38
rs772613159 Chr.1: 11069703-11069703 on Build GRCh38
rs61773683 Chr.1: 11069545-11069545 on Build GRCh38
rs763118096 Chr.1: 11069665-11069666 on Build GRCh38
rs756811037 Chr.1: 11069667-11069667 on Build GRCh38
rs752672802 Chr.1: 11069627-11069627 on Build GRCh38
rs770987829 Chr.1: 11069726-11069726 on Build GRCh38
rs192611779 Chr.1: 11069574-11069574 on Build GRCh38
rs748622049 Chr.1: 11069702-11069702 on Build GRCh38
rs772132007 Chr.1: 11069596-11069596 on Build GRCh38
rs991324336 Chr.1: 11069693-11069693 on Build GRCh38

View Full Product Details