Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50375009-50375280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME1
Entrez Gene ID
146956
Gene Name
essential meiotic structure-specific endonuclease 1
Gene Aliases
MMS4L, SLX2A
UniGene
Hs.514330
Gene Chromosome Location
Chr.17: 50373220-50381479 on Build GRCh38
Transcripts
NM_001166131.1 NM_152463.2 XM_005257081.3 XM_011524395.2 XM_017024236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873739 c.20C>T p.S7L Chr.17: 50375228-50375228 on Build GRCh38
6653653 c.54G>T p.E18D Chr.17: 50375262-50375262 on Build GRCh38
5789110 c.61C>T p.P21S Chr.17: 50375269-50375269 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761429359 Chr.17: 50375203-50375203 on Build GRCh38
rs146276559 Chr.17: 50375246-50375246 on Build GRCh38
rs35248609 Chr.17: 50375223-50375223 on Build GRCh38
rs954675145 Chr.17: 50375075-50375075 on Build GRCh38
rs778157348 Chr.17: 50375255-50375255 on Build GRCh38
rs749620888 Chr.17: 50375256-50375256 on Build GRCh38
rs748401009 Chr.17: 50375251-50375251 on Build GRCh38
rs996422546 Chr.17: 50375055-50375055 on Build GRCh38
rs962883465 Chr.17: 50375108-50375108 on Build GRCh38
rs776322631 Chr.17: 50375201-50375201 on Build GRCh38
rs931265378 Chr.17: 50375200-50375200 on Build GRCh38
rs919853584 Chr.17: 50375195-50375195 on Build GRCh38
rs543903545 Chr.17: 50375215-50375215 on Build GRCh38
rs750149608 Chr.17: 50375214-50375214 on Build GRCh38
rs775021174 Chr.17: 50375189-50375189 on Build GRCh38
rs372340954 Chr.17: 50375196-50375196 on Build GRCh38
rs926760704 Chr.17: 50375125-50375125 on Build GRCh38
rs751542825 Chr.17: 50375227-50375227 on Build GRCh38
rs200348764 Chr.17: 50375184-50375184 on Build GRCh38
rs756225076 Chr.17: 50375253-50375253 on Build GRCh38
rs755067908 Chr.17: 50375230-50375230 on Build GRCh38
rs766253005 Chr.17: 50375224-50375224 on Build GRCh38
rs368491563 Chr.17: 50375192-50375192 on Build GRCh38
rs751505961 Chr.17: 50375159-50375160 on Build GRCh38
rs17714829 Chr.17: 50375120-50375120 on Build GRCh38
rs566019696 Chr.17: 50375043-50375043 on Build GRCh38
rs553645610 Chr.17: 50375059-50375059 on Build GRCh38
rs987989093 Chr.17: 50375088-50375088 on Build GRCh38
rs541428836 Chr.17: 50375130-50375130 on Build GRCh38
rs759519532 Chr.17: 50375258-50375259 on Build GRCh38
rs138848514 Chr.17: 50375156-50375156 on Build GRCh38
rs765009105 Chr.17: 50375209-50375209 on Build GRCh38
rs1029037769 Chr.17: 50375067-50375067 on Build GRCh38

View Full Product Details