Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 89949990-89950261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB11
Entrez Gene ID
90141
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
C14orf143
UniGene
Hs.123232
Gene Chromosome Location
Chr.14: 89794984-89954777 on Build GRCh38
Transcripts
NM_001284266.1 NM_001284267.1 NM_001284268.1 NM_001284269.1 NM_145231.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958588 c.181G>A p.D61N Chr.14: 89950133-89950133 on Build GRCh38
5494689 c.217+4A>C p.? Chr.14: 89950093-89950093 on Build GRCh38
5494690 c.28+4A>C p.? Chr.14: 89950093-89950093 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757619573 Chr.14: 89950185-89950185 on Build GRCh38
rs867724457 Chr.14: 89950127-89950127 on Build GRCh38
rs772886575 Chr.14: 89950141-89950141 on Build GRCh38
rs766970610 Chr.14: 89950136-89950136 on Build GRCh38
rs376114834 Chr.14: 89950170-89950170 on Build GRCh38
rs753124183 Chr.14: 89950171-89950171 on Build GRCh38
rs77780222 Chr.14: 89950173-89950173 on Build GRCh38
rs899592839 Chr.14: 89950152-89950152 on Build GRCh38
rs749234639 Chr.14: 89950101-89950101 on Build GRCh38
rs550304584 Chr.14: 89950111-89950111 on Build GRCh38
rs761342812 Chr.14: 89950129-89950129 on Build GRCh38
rs778001470 Chr.14: 89950048-89950048 on Build GRCh38
rs774173125 Chr.14: 89950124-89950124 on Build GRCh38
rs377273315 Chr.14: 89950088-89950088 on Build GRCh38
rs747224821 Chr.14: 89950055-89950055 on Build GRCh38
rs1021258408 Chr.14: 89950164-89950164 on Build GRCh38
rs780965009 Chr.14: 89950181-89950181 on Build GRCh38
rs992335680 Chr.14: 89950180-89950180 on Build GRCh38
rs371301714 Chr.14: 89950184-89950184 on Build GRCh38
rs749859487 Chr.14: 89950100-89950100 on Build GRCh38
rs766711813 Chr.14: 89950153-89950153 on Build GRCh38
rs769690868 Chr.14: 89950068-89950068 on Build GRCh38
rs760367364 Chr.14: 89950146-89950146 on Build GRCh38
rs763537517 Chr.14: 89950141-89950147 on Build GRCh38
rs777765241 Chr.14: 89950174-89950174 on Build GRCh38
rs879469833 Chr.14: 89950045-89950045 on Build GRCh38
rs768649586 Chr.14: 89950110-89950110 on Build GRCh38
rs371094407 Chr.14: 89950167-89950167 on Build GRCh38
rs755340098 Chr.14: 89950155-89950155 on Build GRCh38
rs1047326281 Chr.14: 89950093-89950093 on Build GRCh38
rs754263776 Chr.14: 89950154-89950154 on Build GRCh38
rs766930860 Chr.14: 89950165-89950166 on Build GRCh38

View Full Product Details