Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147759134-147759359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA5
Entrez Gene ID
2702
Gene Name
gap junction protein alpha 5
Gene Aliases
ATFB11, CX40
UniGene
Hs.447968
Gene Chromosome Location
Chr.1: 147756199-147781127 on Build GRCh38
Transcripts
NM_005266.6 NM_181703.3 XM_005272951.4 XM_017001044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5421586 c.15C>A p.S5R Chr.1: 147759224-147759224 on Build GRCh38
895329 c.42A>G p.V14V Chr.1: 147759197-147759197 on Build GRCh38
5377528 c.57C>T p.T19T Chr.1: 147759182-147759182 on Build GRCh38
2121840 c.30C>T p.F10F Chr.1: 147759209-147759209 on Build GRCh38
139314 c.97C>T p.R33C Chr.1: 147759142-147759142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782751879 Chr.1: 147759167-147759167 on Build GRCh38
rs981040058 Chr.1: 147759170-147759170 on Build GRCh38
rs373059679 Chr.1: 147759198-147759198 on Build GRCh38
rs781923934 Chr.1: 147759285-147759285 on Build GRCh38
rs782282057 Chr.1: 147759275-147759275 on Build GRCh38
rs781784744 Chr.1: 147759215-147759215 on Build GRCh38
rs782806068 Chr.1: 147759203-147759203 on Build GRCh38
rs2232190 Chr.1: 147759265-147759265 on Build GRCh38
rs781879341 Chr.1: 147759262-147759262 on Build GRCh38
rs888796876 Chr.1: 147759310-147759310 on Build GRCh38
rs782228278 Chr.1: 147759237-147759237 on Build GRCh38
rs1007411229 Chr.1: 147759314-147759314 on Build GRCh38
rs782200107 Chr.1: 147759286-147759286 on Build GRCh38
rs782419411 Chr.1: 147759280-147759280 on Build GRCh38
rs782537233 Chr.1: 147759186-147759186 on Build GRCh38
rs587770589 Chr.1: 147759230-147759230 on Build GRCh38
rs140502327 Chr.1: 147759253-147759253 on Build GRCh38
rs1043368131 Chr.1: 147759305-147759305 on Build GRCh38
rs144069395 Chr.1: 147759226-147759226 on Build GRCh38

View Full Product Details