Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18398444-18398717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZANK1
Entrez Gene ID
55184
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
UniGene
Hs.472225
Gene Chromosome Location
Chr.20: 18383367-18467185 on Build GRCh38
Transcripts
NM_001099407.1 XM_005260742.3 XM_005260743.3 XM_006723584.2 XM_011529268.2 XM_011529271.2 XM_011529272.2 XM_011529275.2 XM_011529277.2 XM_017027909.1 XM_017027910.1 XM_017027911.1 XM_017027912.1 XM_017027913.1 XM_017027914.1 XM_017027915.1 XM_017027916.1 XM_017027917.1 XM_017027918.1 XM_017027919.1 XM_017027920.1 XM_017027921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448912 c.1433-2A>G p.? Chr.20: 18398628-18398628 on Build GRCh38
7164297 c.1524A>G p.A508A Chr.20: 18398541-18398541 on Build GRCh38
5018633 c.1166C>T p.A389V Chr.20: 18398551-18398551 on Build GRCh38
5952432 c.1516G>A p.A506T Chr.20: 18398543-18398543 on Build GRCh38
1024875 c.1504C>T p.R502W Chr.20: 18398555-18398555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777443153 Chr.20: 18398568-18398568 on Build GRCh38
rs1002367097 Chr.20: 18398620-18398620 on Build GRCh38
rs574023983 Chr.20: 18398554-18398554 on Build GRCh38
rs768860531 Chr.20: 18398597-18398597 on Build GRCh38
rs766832289 Chr.20: 18398551-18398551 on Build GRCh38
rs200789965 Chr.20: 18398545-18398545 on Build GRCh38
rs760784763 Chr.20: 18398658-18398658 on Build GRCh38
rs760087765 Chr.20: 18398670-18398670 on Build GRCh38
rs745687904 Chr.20: 18398494-18398494 on Build GRCh38
rs772233794 Chr.20: 18398647-18398647 on Build GRCh38
rs372483532 Chr.20: 18398486-18398486 on Build GRCh38
rs777064640 Chr.20: 18398667-18398667 on Build GRCh38
rs755349093 Chr.20: 18398555-18398555 on Build GRCh38
rs143846023 Chr.20: 18398485-18398485 on Build GRCh38
rs752362705 Chr.20: 18398565-18398565 on Build GRCh38
rs562949522 Chr.20: 18398681-18398681 on Build GRCh38
rs769542582 Chr.20: 18398629-18398629 on Build GRCh38
rs774493084 Chr.20: 18398503-18398503 on Build GRCh38
rs781037965 Chr.20: 18398594-18398594 on Build GRCh38
rs766463003 Chr.20: 18398665-18398665 on Build GRCh38
rs577908080 Chr.20: 18398648-18398648 on Build GRCh38
rs915058335 Chr.20: 18398611-18398611 on Build GRCh38
rs769607318 Chr.20: 18398498-18398498 on Build GRCh38
rs201365109 Chr.20: 18398590-18398590 on Build GRCh38
rs376467555 Chr.20: 18398491-18398491 on Build GRCh38
rs777421241 Chr.20: 18398474-18398474 on Build GRCh38
rs767588397 Chr.20: 18398540-18398540 on Build GRCh38
rs756577408 Chr.20: 18398633-18398635 on Build GRCh38
rs761193282 Chr.20: 18398550-18398550 on Build GRCh38
rs181700509 Chr.20: 18398578-18398578 on Build GRCh38
rs952954042 Chr.20: 18398678-18398678 on Build GRCh38
rs374370227 Chr.20: 18398583-18398583 on Build GRCh38
rs148616789 Chr.20: 18398672-18398672 on Build GRCh38
rs748382947 Chr.20: 18398646-18398646 on Build GRCh38
rs758061055 Chr.20: 18398566-18398566 on Build GRCh38
rs745802938 Chr.20: 18398596-18398596 on Build GRCh38
rs762017045 Chr.20: 18398508-18398508 on Build GRCh38
rs775288141 Chr.20: 18398638-18398638 on Build GRCh38
rs6111938 Chr.20: 18398609-18398609 on Build GRCh38
rs555802178 Chr.20: 18398478-18398478 on Build GRCh38
rs1032137962 Chr.20: 18398506-18398506 on Build GRCh38

View Full Product Details