Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86266110-86266345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EED
Entrez Gene ID
8726
Gene Name
embryonic ectoderm development
Gene Aliases
HEED, WAIT1
UniGene
Hs.503510
Gene Chromosome Location
Chr.11: 86244384-86285420 on Build GRCh38
Transcripts
NM_001308007.1 NM_003797.4 XM_005274373.2 XM_011545330.1 XM_011545331.1 XM_017018512.1 XM_017018513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
691224 c.788G>T p.W263L Chr.11: 86266144-86266144 on Build GRCh38
3792004 c.778C>T p.L260F Chr.11: 86266134-86266134 on Build GRCh38
4507840 c.758C>T p.S253F Chr.11: 86266114-86266114 on Build GRCh38
6365110 c.793A>G p.I265V Chr.11: 86266149-86266149 on Build GRCh38
6935405 c.791G>T p.R264M Chr.11: 86266147-86266147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770256429 Chr.11: 86266308-86266308 on Build GRCh38
rs777218182 Chr.11: 86266223-86266223 on Build GRCh38
rs201730128 Chr.11: 86266227-86266227 on Build GRCh38
rs573849181 Chr.11: 86266293-86266293 on Build GRCh38
rs562701665 Chr.11: 86266302-86266302 on Build GRCh38
rs367891435 Chr.11: 86266176-86266176 on Build GRCh38
rs891764147 Chr.11: 86266235-86266235 on Build GRCh38
rs374042174 Chr.11: 86266289-86266289 on Build GRCh38
rs374311051 Chr.11: 86266153-86266153 on Build GRCh38
rs1018181045 Chr.11: 86266294-86266294 on Build GRCh38
rs953960092 Chr.11: 86266296-86266296 on Build GRCh38
rs905981541 Chr.11: 86266166-86266166 on Build GRCh38
rs542865901 Chr.11: 86266295-86266295 on Build GRCh38
rs1021790756 Chr.11: 86266265-86266265 on Build GRCh38
rs1042949721 Chr.11: 86266248-86266248 on Build GRCh38
rs755834675 Chr.11: 86266258-86266258 on Build GRCh38
rs779476300 Chr.11: 86266267-86266267 on Build GRCh38
rs767351539 Chr.11: 86266247-86266247 on Build GRCh38
rs945903210 Chr.11: 86266246-86266246 on Build GRCh38
rs763956385 Chr.11: 86266169-86266169 on Build GRCh38
rs775716311 Chr.11: 86266147-86266147 on Build GRCh38
rs560002956 Chr.11: 86266238-86266238 on Build GRCh38
rs938759887 Chr.11: 86266171-86266171 on Build GRCh38
rs750065638 Chr.11: 86266256-86266256 on Build GRCh38

View Full Product Details