Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15355167-15355394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGF
Entrez Gene ID
2277
Gene Name
c-fos induced growth factor
Gene Aliases
VEGF-D, VEGFD
UniGene
Hs.11392
Gene Chromosome Location
Chr.X: 15345591-15384413 on Build GRCh38
Transcripts
NM_004469.4 NR_037859.2

Gene Symbol
PIR-FIGF
Entrez Gene ID
100532742
Gene Name
PIR-FIGF readthrough
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15345591-15491309 on Build GRCh38
Transcripts
NM_004469.4 NR_037859.2

SNP Detail

SNP ID Chromosome Location
rs750311027 Chr.X: 15355322-15355322 on Build GRCh38
rs775313766 Chr.X: 15355235-15355235 on Build GRCh38
rs4531212 Chr.X: 15355347-15355347 on Build GRCh38
rs149425095 Chr.X: 15355260-15355260 on Build GRCh38
rs773703171 Chr.X: 15355348-15355350 on Build GRCh38
rs750973484 Chr.X: 15355363-15355363 on Build GRCh38
rs768262468 Chr.X: 15355344-15355346 on Build GRCh38
rs376317191 Chr.X: 15355198-15355198 on Build GRCh38
rs771466661 Chr.X: 15355210-15355210 on Build GRCh38
rs769643857 Chr.X: 15355335-15355336 on Build GRCh38
rs748325706 Chr.X: 15355201-15355201 on Build GRCh38
rs778064277 Chr.X: 15355348-15355348 on Build GRCh38
rs759134392 Chr.X: 15355211-15355211 on Build GRCh38
rs756343010 Chr.X: 15355346-15355346 on Build GRCh38
rs752375557 Chr.X: 15355329-15355329 on Build GRCh38
rs776474991 Chr.X: 15355277-15355277 on Build GRCh38
rs748843265 Chr.X: 15355343-15355344 on Build GRCh38
rs369324337 Chr.X: 15355203-15355203 on Build GRCh38
rs149259363 Chr.X: 15355263-15355263 on Build GRCh38
rs960098556 Chr.X: 15355367-15355367 on Build GRCh38
rs183118910 Chr.X: 15355287-15355287 on Build GRCh38
rs765232774 Chr.X: 15355288-15355288 on Build GRCh38
rs775238907 Chr.X: 15355336-15355339 on Build GRCh38
rs1037986623 Chr.X: 15355290-15355290 on Build GRCh38
rs755080879 Chr.X: 15355328-15355328 on Build GRCh38
rs771663218 Chr.X: 15355233-15355233 on Build GRCh38
rs143510014 Chr.X: 15355270-15355270 on Build GRCh38
rs761445938 Chr.X: 15355280-15355280 on Build GRCh38
rs187053601 Chr.X: 15355332-15355332 on Build GRCh38
rs1020187289 Chr.X: 15355237-15355237 on Build GRCh38
rs778173575 Chr.X: 15355202-15355202 on Build GRCh38

View Full Product Details