Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68464737-68464998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO48
Entrez Gene ID
554251
Gene Name
F-box protein 48
Gene Aliases
-
UniGene
Hs.164117
Gene Chromosome Location
Chr.2: 68459419-68467325 on Build GRCh38
Transcripts
NM_001024680.2 XM_005264407.2 XM_017004437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661824 c.216T>G p.S72R Chr.2: 68464930-68464930 on Build GRCh38
5474501 c.185G>A p.C62Y Chr.2: 68464961-68464961 on Build GRCh38
7158560 c.263G>C p.R88P Chr.2: 68464883-68464883 on Build GRCh38
5059014 c.204A>G p.T68T Chr.2: 68464942-68464942 on Build GRCh38
5397229 c.262C>T p.R88* Chr.2: 68464884-68464884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369259812 Chr.2: 68464949-68464949 on Build GRCh38
rs780895382 Chr.2: 68464842-68464842 on Build GRCh38
rs1018603408 Chr.2: 68464766-68464766 on Build GRCh38
rs762821956 Chr.2: 68464941-68464941 on Build GRCh38
rs778019907 Chr.2: 68464937-68464940 on Build GRCh38
rs892405836 Chr.2: 68464770-68464770 on Build GRCh38
rs751240892 Chr.2: 68464825-68464825 on Build GRCh38
rs566301181 Chr.2: 68464819-68464819 on Build GRCh38
rs746274887 Chr.2: 68464843-68464843 on Build GRCh38
rs768860184 Chr.2: 68464865-68464865 on Build GRCh38
rs1022607264 Chr.2: 68464781-68464781 on Build GRCh38
rs1009499270 Chr.2: 68464777-68464777 on Build GRCh38
rs920133176 Chr.2: 68464974-68464974 on Build GRCh38
rs200307344 Chr.2: 68464883-68464883 on Build GRCh38
rs139903910 Chr.2: 68464872-68464872 on Build GRCh38
rs756762751 Chr.2: 68464955-68464955 on Build GRCh38
rs1037424906 Chr.2: 68464787-68464787 on Build GRCh38
rs773448216 Chr.2: 68464905-68464905 on Build GRCh38
rs368567725 Chr.2: 68464823-68464823 on Build GRCh38
rs148436824 Chr.2: 68464963-68464963 on Build GRCh38
rs759372116 Chr.2: 68464908-68464908 on Build GRCh38
rs765194890 Chr.2: 68464820-68464820 on Build GRCh38
rs374687975 Chr.2: 68464793-68464793 on Build GRCh38
rs780728736 Chr.2: 68464853-68464853 on Build GRCh38
rs908528303 Chr.2: 68464922-68464922 on Build GRCh38
rs941811846 Chr.2: 68464782-68464782 on Build GRCh38
rs748243476 Chr.2: 68464886-68464886 on Build GRCh38
rs750047960 Chr.2: 68464977-68464977 on Build GRCh38
rs775414637 Chr.2: 68464937-68464937 on Build GRCh38
rs1002378411 Chr.2: 68464807-68464807 on Build GRCh38
rs756526283 Chr.2: 68464845-68464845 on Build GRCh38
rs896773578 Chr.2: 68464837-68464837 on Build GRCh38
rs145911210 Chr.2: 68464801-68464801 on Build GRCh38
rs764980463 Chr.2: 68464927-68464927 on Build GRCh38
rs760943896 Chr.2: 68464796-68464796 on Build GRCh38
rs749772157 Chr.2: 68464857-68464857 on Build GRCh38
rs772246905 Chr.2: 68464895-68464895 on Build GRCh38
rs202123162 Chr.2: 68464822-68464822 on Build GRCh38
rs557893848 Chr.2: 68464942-68464942 on Build GRCh38
rs756921216 Chr.2: 68464841-68464841 on Build GRCh38

View Full Product Details