Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69574473-69574738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS2
Entrez Gene ID
10818
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
UniGene
Hs.593446
Gene Chromosome Location
Chr.12: 69470349-69579793 on Build GRCh38
Transcripts
NM_001042555.2 NM_001278351.1 NM_001278353.1 NM_001278354.1 NM_001278355.1 NM_001278356.1 NM_001278357.1 NM_006654.4 XM_017018717.1 XM_017018718.1 XM_017018719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6294193 c.1277G>T p.R426L Chr.12: 69574705-69574705 on Build GRCh38
4835499 c.1288G>A p.E430K Chr.12: 69574716-69574716 on Build GRCh38
7368836 c.1216A>G p.K406E Chr.12: 69574644-69574644 on Build GRCh38
6137802 c.1239G>T p.R413R Chr.12: 69574667-69574667 on Build GRCh38
7316017 c.1135C>G p.L379V Chr.12: 69574563-69574563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370860144 Chr.12: 69574685-69574685 on Build GRCh38
rs768688491 Chr.12: 69574528-69574528 on Build GRCh38
rs370771368 Chr.12: 69574637-69574637 on Build GRCh38
rs375527184 Chr.12: 69574591-69574591 on Build GRCh38
rs377368151 Chr.12: 69574675-69574675 on Build GRCh38
rs375588784 Chr.12: 69574704-69574704 on Build GRCh38
rs758123047 Chr.12: 69574682-69574682 on Build GRCh38
rs187299511 Chr.12: 69574512-69574512 on Build GRCh38
rs377766786 Chr.12: 69574708-69574708 on Build GRCh38
rs769656225 Chr.12: 69574659-69574659 on Build GRCh38
rs200715711 Chr.12: 69574572-69574572 on Build GRCh38
rs778440805 Chr.12: 69574593-69574593 on Build GRCh38
rs751184690 Chr.12: 69574694-69574694 on Build GRCh38
rs761048105 Chr.12: 69574678-69574678 on Build GRCh38
rs112443855 Chr.12: 69574665-69574665 on Build GRCh38
rs1013221115 Chr.12: 69574551-69574551 on Build GRCh38
rs747962982 Chr.12: 69574653-69574653 on Build GRCh38
rs749215723 Chr.12: 69574605-69574605 on Build GRCh38
rs767997912 Chr.12: 69574584-69574584 on Build GRCh38
rs781293067 Chr.12: 69574706-69574706 on Build GRCh38
rs777150070 Chr.12: 69574666-69574666 on Build GRCh38
rs770210154 Chr.12: 69574559-69574559 on Build GRCh38
rs752037897 Chr.12: 69574576-69574576 on Build GRCh38
rs766434579 Chr.12: 69574575-69574575 on Build GRCh38
rs756403029 Chr.12: 69574592-69574592 on Build GRCh38
rs369512504 Chr.12: 69574504-69574504 on Build GRCh38
rs369774666 Chr.12: 69574610-69574610 on Build GRCh38
rs1034730582 Chr.12: 69574518-69574518 on Build GRCh38
rs761689738 Chr.12: 69574547-69574547 on Build GRCh38
rs376497327 Chr.12: 69574502-69574502 on Build GRCh38
rs372662720 Chr.12: 69574630-69574630 on Build GRCh38
rs764475417 Chr.12: 69574680-69574680 on Build GRCh38
rs756070543 Chr.12: 69574555-69574555 on Build GRCh38
rs189315944 Chr.12: 69574674-69574674 on Build GRCh38
rs781274920 Chr.12: 69574649-69574649 on Build GRCh38
rs977611851 Chr.12: 69574643-69574643 on Build GRCh38
rs945337284 Chr.12: 69574705-69574705 on Build GRCh38
rs760085317 Chr.12: 69574581-69574581 on Build GRCh38
rs773329122 Chr.12: 69574662-69574662 on Build GRCh38
rs762956505 Chr.12: 69574573-69574573 on Build GRCh38
rs775308985 Chr.12: 69574513-69574513 on Build GRCh38
rs772139122 Chr.12: 69574688-69574688 on Build GRCh38
rs746921030 Chr.12: 69574526-69574526 on Build GRCh38
rs34097404 Chr.12: 69574663-69574663 on Build GRCh38
rs376137375 Chr.12: 69574631-69574631 on Build GRCh38
rs200964541 Chr.12: 69574671-69574671 on Build GRCh38
rs759605335 Chr.12: 69574670-69574670 on Build GRCh38
rs776621252 Chr.12: 69574531-69574531 on Build GRCh38
rs779659621 Chr.12: 69574604-69574604 on Build GRCh38
rs567234657 Chr.12: 69574667-69574667 on Build GRCh38
rs760336153 Chr.12: 69574681-69574681 on Build GRCh38
rs757873308 Chr.12: 69574600-69574600 on Build GRCh38

View Full Product Details