Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47725933-47726204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD2
Entrez Gene ID
30846
Gene Name
EH domain containing 2
Gene Aliases
PAST2
UniGene
Hs.744963
Gene Chromosome Location
Chr.19: 47713344-47743134 on Build GRCh38
Transcripts
NM_014601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998788 c.716C>T p.A239V Chr.19: 47726025-47726025 on Build GRCh38
998789 c.752G>A p.R251H Chr.19: 47726061-47726061 on Build GRCh38
7365992 c.704C>T p.A235V Chr.19: 47726013-47726013 on Build GRCh38
4079767 c.754G>A p.V252I Chr.19: 47726063-47726063 on Build GRCh38
3106690 c.802C>T p.R268W Chr.19: 47726111-47726111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948649576 Chr.19: 47725984-47725984 on Build GRCh38
rs1025530886 Chr.19: 47726014-47726014 on Build GRCh38
rs773625785 Chr.19: 47726166-47726166 on Build GRCh38
rs544914999 Chr.19: 47726011-47726011 on Build GRCh38
rs757181096 Chr.19: 47725976-47725976 on Build GRCh38
rs150300804 Chr.19: 47726147-47726147 on Build GRCh38
rs1006684929 Chr.19: 47725985-47725985 on Build GRCh38
rs199961822 Chr.19: 47726152-47726152 on Build GRCh38
rs752344653 Chr.19: 47726102-47726102 on Build GRCh38
rs984101500 Chr.19: 47726084-47726084 on Build GRCh38
rs755011408 Chr.19: 47726141-47726141 on Build GRCh38
rs771422226 Chr.19: 47726169-47726169 on Build GRCh38
rs763918440 Chr.19: 47726109-47726109 on Build GRCh38
rs969536924 Chr.19: 47726138-47726138 on Build GRCh38
rs781570240 Chr.19: 47726007-47726007 on Build GRCh38
rs767163876 Chr.19: 47726098-47726098 on Build GRCh38
rs367574070 Chr.19: 47725986-47725986 on Build GRCh38
rs757175078 Chr.19: 47726114-47726114 on Build GRCh38
rs775008579 Chr.19: 47726071-47726071 on Build GRCh38
rs547172741 Chr.19: 47726076-47726076 on Build GRCh38
rs749553791 Chr.19: 47726168-47726168 on Build GRCh38
rs996023845 Chr.19: 47726096-47726096 on Build GRCh38
rs928139468 Chr.19: 47726160-47726160 on Build GRCh38
rs748350898 Chr.19: 47726018-47726018 on Build GRCh38
rs774957397 Chr.19: 47726183-47726183 on Build GRCh38
rs951479746 Chr.19: 47726020-47726020 on Build GRCh38
rs747150177 Chr.19: 47725996-47725996 on Build GRCh38
rs560354382 Chr.19: 47726111-47726111 on Build GRCh38
rs576144192 Chr.19: 47726008-47726008 on Build GRCh38
rs767063664 Chr.19: 47726063-47726063 on Build GRCh38
rs755769922 Chr.19: 47726104-47726104 on Build GRCh38
rs533522326 Chr.19: 47726062-47726062 on Build GRCh38
rs781274712 Chr.19: 47726146-47726146 on Build GRCh38
rs749225110 Chr.19: 47725966-47725966 on Build GRCh38
rs769953115 Chr.19: 47726159-47726159 on Build GRCh38
rs778765086 Chr.19: 47726122-47726122 on Build GRCh38
rs1028472701 Chr.19: 47726099-47726099 on Build GRCh38
rs770150064 Chr.19: 47726035-47726035 on Build GRCh38
rs867611192 Chr.19: 47726126-47726126 on Build GRCh38
rs564807856 Chr.19: 47726056-47726056 on Build GRCh38
rs745812114 Chr.19: 47726132-47726132 on Build GRCh38
rs1019831845 Chr.19: 47726158-47726158 on Build GRCh38
rs1043214730 Chr.19: 47725954-47725954 on Build GRCh38
rs777450158 Chr.19: 47725955-47725955 on Build GRCh38

View Full Product Details