Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35279899-35280150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF6
Entrez Gene ID
3692
Gene Name
eukaryotic translation initiation factor 6
Gene Aliases
CAB, EIF3A, ITGB4BP, b(2)gcn, eIF-6, p27(BBP), p27BBP
UniGene
Hs.743992
Gene Chromosome Location
Chr.20: 35278906-35284816 on Build GRCh38
Transcripts
NM_001267810.1 NM_002212.3 NM_181466.2 NM_181468.2 NR_052022.1 NR_052023.1 NR_052024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1411390 c.427G>T p.A143S Chr.20: 35280061-35280061 on Build GRCh38
4545364 c.319G>A p.E107K Chr.20: 35280112-35280112 on Build GRCh38
1326789 c.379G>C p.E127Q Chr.20: 35280109-35280109 on Build GRCh38
232241 c.539C>T p.P180L Chr.20: 35279949-35279949 on Build GRCh38
6159893 c.379G>A p.E127K Chr.20: 35280109-35280109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879141800 Chr.20: 35280117-35280118 on Build GRCh38
rs750826735 Chr.20: 35280087-35280087 on Build GRCh38
rs866211331 Chr.20: 35280059-35280059 on Build GRCh38
rs778932456 Chr.20: 35280124-35280124 on Build GRCh38
rs756430831 Chr.20: 35280091-35280091 on Build GRCh38
rs77914324 Chr.20: 35279951-35279951 on Build GRCh38
rs770495030 Chr.20: 35280014-35280014 on Build GRCh38
rs760487812 Chr.20: 35279988-35279988 on Build GRCh38
rs2250205 Chr.20: 35280121-35280121 on Build GRCh38
rs895428184 Chr.20: 35280105-35280105 on Build GRCh38
rs934143137 Chr.20: 35279927-35279927 on Build GRCh38
rs764785068 Chr.20: 35280057-35280057 on Build GRCh38
rs768741006 Chr.20: 35279977-35279977 on Build GRCh38
rs758995002 Chr.20: 35280026-35280026 on Build GRCh38
rs773204593 Chr.20: 35279960-35279960 on Build GRCh38
rs772980128 Chr.20: 35279999-35279999 on Build GRCh38
rs780261771 Chr.20: 35279959-35279959 on Build GRCh38
rs370783372 Chr.20: 35279916-35279916 on Build GRCh38
rs374147852 Chr.20: 35280021-35280021 on Build GRCh38
rs748054112 Chr.20: 35279989-35279989 on Build GRCh38
rs11553027 Chr.20: 35280016-35280016 on Build GRCh38
rs778014353 Chr.20: 35279971-35279971 on Build GRCh38
rs771923186 Chr.20: 35279997-35279997 on Build GRCh38
rs768049543 Chr.20: 35280060-35280060 on Build GRCh38
rs766540370 Chr.20: 35280100-35280100 on Build GRCh38
rs781660023 Chr.20: 35279938-35279938 on Build GRCh38
rs749332022 Chr.20: 35279966-35279966 on Build GRCh38
rs1802240 Chr.20: 35279969-35279969 on Build GRCh38
rs144752898 Chr.20: 35280058-35280058 on Build GRCh38
rs756273702 Chr.20: 35279954-35279954 on Build GRCh38
rs760460233 Chr.20: 35280000-35280000 on Build GRCh38
rs757748212 Chr.20: 35279931-35279931 on Build GRCh38
rs754043227 Chr.20: 35280117-35280117 on Build GRCh38
rs1006354941 Chr.20: 35280073-35280073 on Build GRCh38
rs752066001 Chr.20: 35279918-35279918 on Build GRCh38
rs770733564 Chr.20: 35280113-35280113 on Build GRCh38

View Full Product Details