Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124831294-124831565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175B
Entrez Gene ID
23172
Gene Name
family with sequence similarity 175 member B
Gene Aliases
ABRO1, KIAA0157
UniGene
Hs.280695
Gene Chromosome Location
Chr.10: 124801785-124836670 on Build GRCh38
Transcripts
NM_032182.3 XM_011539554.1 XM_017015986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978218 c.695G>A p.R232Q Chr.10: 124831380-124831380 on Build GRCh38
256906 c.696A>G p.R232R Chr.10: 124831381-124831381 on Build GRCh38
4790140 c.709T>C p.C237R Chr.10: 124831394-124831394 on Build GRCh38
1193301 c.671G>A p.C224Y Chr.10: 124831356-124831356 on Build GRCh38
1346586 c.776G>T p.R259I Chr.10: 124831461-124831461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773670652 Chr.10: 124831395-124831395 on Build GRCh38
rs770682699 Chr.10: 124831455-124831458 on Build GRCh38
rs746978901 Chr.10: 124831344-124831344 on Build GRCh38
rs73369372 Chr.10: 124831476-124831476 on Build GRCh38
rs369758673 Chr.10: 124831478-124831478 on Build GRCh38
rs200885996 Chr.10: 124831374-124831374 on Build GRCh38
rs780989802 Chr.10: 124831352-124831352 on Build GRCh38
rs1033224859 Chr.10: 124831525-124831525 on Build GRCh38
rs553687640 Chr.10: 124831544-124831544 on Build GRCh38
rs761368917 Chr.10: 124831535-124831535 on Build GRCh38
rs373070820 Chr.10: 124831391-124831391 on Build GRCh38
rs761086489 Chr.10: 124831443-124831443 on Build GRCh38
rs866654451 Chr.10: 124831379-124831379 on Build GRCh38
rs752677538 Chr.10: 124831486-124831486 on Build GRCh38
rs535344186 Chr.10: 124831524-124831524 on Build GRCh38
rs772218239 Chr.10: 124831329-124831331 on Build GRCh38
rs1050271084 Chr.10: 124831420-124831420 on Build GRCh38
rs369077371 Chr.10: 124831382-124831382 on Build GRCh38
rs768086145 Chr.10: 124831402-124831402 on Build GRCh38
rs537159917 Chr.10: 124831461-124831461 on Build GRCh38
rs755076386 Chr.10: 124831329-124831329 on Build GRCh38
rs959151351 Chr.10: 124831533-124831533 on Build GRCh38
rs745730498 Chr.10: 124831499-124831499 on Build GRCh38
rs773757845 Chr.10: 124831442-124831442 on Build GRCh38
rs781077918 Chr.10: 124831494-124831494 on Build GRCh38
rs745642408 Chr.10: 124831354-124831354 on Build GRCh38
rs766770349 Chr.10: 124831459-124831459 on Build GRCh38
rs368433044 Chr.10: 124831500-124831500 on Build GRCh38
rs748810930 Chr.10: 124831383-124831383 on Build GRCh38
rs754007696 Chr.10: 124831323-124831323 on Build GRCh38
rs762415831 Chr.10: 124831400-124831400 on Build GRCh38
rs757270314 Chr.10: 124831488-124831488 on Build GRCh38
rs770705420 Chr.10: 124831346-124831346 on Build GRCh38
rs36120522 Chr.10: 124831500-124831501 on Build GRCh38
rs769477930 Chr.10: 124831375-124831375 on Build GRCh38
rs746858225 Chr.10: 124831444-124831445 on Build GRCh38
rs1000613064 Chr.10: 124831503-124831503 on Build GRCh38
rs375402637 Chr.10: 124831328-124831328 on Build GRCh38
rs773437994 Chr.10: 124831442-124831443 on Build GRCh38
rs777629929 Chr.10: 124831332-124831332 on Build GRCh38
rs59157327 Chr.10: 124831348-124831348 on Build GRCh38

View Full Product Details