Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38686803-38687062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLO1
Entrez Gene ID
2739
Gene Name
glyoxalase I
Gene Aliases
GLOD1, GLYI, HEL-S-74
UniGene
Hs.268849
Gene Chromosome Location
Chr.6: 38675925-38703176 on Build GRCh38
Transcripts
NM_006708.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208318 c.167+1G>A p.? Chr.6: 38686891-38686891 on Build GRCh38
4445057 c.167C>T p.T56M Chr.6: 38686892-38686892 on Build GRCh38
3076855 c.112C>T p.R38* Chr.6: 38686947-38686947 on Build GRCh38
5000484 c.100C>T p.Q34* Chr.6: 38686959-38686959 on Build GRCh38
1078746 c.150T>C p.T50T Chr.6: 38686909-38686909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560135215 Chr.6: 38686908-38686908 on Build GRCh38
rs754689081 Chr.6: 38687010-38687010 on Build GRCh38
rs764033567 Chr.6: 38686852-38686852 on Build GRCh38
rs755657471 Chr.6: 38686883-38686883 on Build GRCh38
rs749861545 Chr.6: 38686975-38686975 on Build GRCh38
rs778253594 Chr.6: 38686889-38686890 on Build GRCh38
rs936450419 Chr.6: 38686931-38686931 on Build GRCh38
rs761785417 Chr.6: 38686971-38686971 on Build GRCh38
rs781072792 Chr.6: 38686918-38686918 on Build GRCh38
rs760253626 Chr.6: 38686984-38686984 on Build GRCh38
rs142451043 Chr.6: 38686862-38686862 on Build GRCh38
rs749567329 Chr.6: 38686891-38686891 on Build GRCh38
rs372860215 Chr.6: 38686957-38686957 on Build GRCh38
rs4451130 Chr.6: 38686968-38686968 on Build GRCh38
rs376365555 Chr.6: 38687019-38687019 on Build GRCh38
rs199584798 Chr.6: 38686871-38686871 on Build GRCh38
rs761910099 Chr.6: 38686864-38686864 on Build GRCh38
rs1937775 Chr.6: 38686835-38686835 on Build GRCh38
rs768570333 Chr.6: 38686960-38686960 on Build GRCh38
rs774346577 Chr.6: 38686969-38686969 on Build GRCh38
rs758740372 Chr.6: 38686902-38686902 on Build GRCh38
rs900212618 Chr.6: 38686966-38686966 on Build GRCh38
rs143771246 Chr.6: 38686959-38686959 on Build GRCh38
rs139437714 Chr.6: 38686947-38686947 on Build GRCh38
rs767497323 Chr.6: 38686974-38686974 on Build GRCh38
rs150725457 Chr.6: 38686892-38686892 on Build GRCh38
rs778094568 Chr.6: 38687012-38687012 on Build GRCh38
rs749964753 Chr.6: 38686875-38686875 on Build GRCh38
rs765854991 Chr.6: 38686992-38686992 on Build GRCh38
rs1038747749 Chr.6: 38686946-38686946 on Build GRCh38
rs771521865 Chr.6: 38686909-38686909 on Build GRCh38
rs796746763 Chr.6: 38686954-38686954 on Build GRCh38
rs769031846 Chr.6: 38686905-38686905 on Build GRCh38
rs745962006 Chr.6: 38686920-38686920 on Build GRCh38
rs753455968 Chr.6: 38687007-38687007 on Build GRCh38

View Full Product Details