Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89190245-89190474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP4
Entrez Gene ID
115361
Gene Name
guanylate binding protein 4
Gene Aliases
Mpa2
UniGene
Hs.409925
Gene Chromosome Location
Chr.1: 89181148-89198950 on Build GRCh38
Transcripts
NM_052941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6494366 c.924G>A p.G308G Chr.1: 89190311-89190311 on Build GRCh38
4636981 c.919C>A p.L307M Chr.1: 89190316-89190316 on Build GRCh38
5934304 c.917-3C>T p.? Chr.1: 89190321-89190321 on Build GRCh38
6127921 c.986C>T p.A329V Chr.1: 89190249-89190249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757537588 Chr.1: 89190284-89190284 on Build GRCh38
rs755443191 Chr.1: 89190330-89190330 on Build GRCh38
rs111594101 Chr.1: 89190278-89190278 on Build GRCh38
rs1410871 Chr.1: 89190311-89190311 on Build GRCh38
rs542414272 Chr.1: 89190363-89190363 on Build GRCh38
rs201574307 Chr.1: 89190299-89190299 on Build GRCh38
rs371314987 Chr.1: 89190293-89190293 on Build GRCh38
rs769828427 Chr.1: 89190278-89190278 on Build GRCh38
rs778388918 Chr.1: 89190338-89190338 on Build GRCh38
rs1045473554 Chr.1: 89190392-89190392 on Build GRCh38
rs934422941 Chr.1: 89190417-89190417 on Build GRCh38
rs759454847 Chr.1: 89190364-89190364 on Build GRCh38
rs150829936 Chr.1: 89190276-89190276 on Build GRCh38
rs577917792 Chr.1: 89190448-89190450 on Build GRCh38
rs771692300 Chr.1: 89190347-89190347 on Build GRCh38
rs746672685 Chr.1: 89190354-89190354 on Build GRCh38
rs948392838 Chr.1: 89190388-89190388 on Build GRCh38
rs200509294 Chr.1: 89190349-89190349 on Build GRCh38
rs747716386 Chr.1: 89190344-89190344 on Build GRCh38
rs906950530 Chr.1: 89190404-89190405 on Build GRCh38
rs749744566 Chr.1: 89190326-89190326 on Build GRCh38
rs928540528 Chr.1: 89190281-89190281 on Build GRCh38
rs543748146 Chr.1: 89190444-89190444 on Build GRCh38
rs770714474 Chr.1: 89190360-89190360 on Build GRCh38
rs560707763 Chr.1: 89190373-89190373 on Build GRCh38
rs780304326 Chr.1: 89190317-89190317 on Build GRCh38
rs374274451 Chr.1: 89190272-89190272 on Build GRCh38
rs12038448 Chr.1: 89190319-89190319 on Build GRCh38
rs10922590 Chr.1: 89190440-89190440 on Build GRCh38

View Full Product Details