Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5394602-5394875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4810564 c.1118C>T p.S373L Chr.18: 5394744-5394744 on Build GRCh38
4681175 c.3209C>T p.T1070I Chr.18: 5394738-5394738 on Build GRCh38
1303779 c.3203C>T p.S1068L Chr.18: 5394744-5394744 on Build GRCh38
7255387 c.1075G>A p.A359T Chr.18: 5394787-5394787 on Build GRCh38
3989594 c.3182A>C p.K1061T Chr.18: 5394765-5394765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776396188 Chr.18: 5394650-5394650 on Build GRCh38
rs777246251 Chr.18: 5394700-5394700 on Build GRCh38
rs773272519 Chr.18: 5394796-5394796 on Build GRCh38
rs976801568 Chr.18: 5394808-5394808 on Build GRCh38
rs778843417 Chr.18: 5394742-5394742 on Build GRCh38
rs199831654 Chr.18: 5394680-5394680 on Build GRCh38
rs767551000 Chr.18: 5394793-5394793 on Build GRCh38
rs761964315 Chr.18: 5394792-5394792 on Build GRCh38
rs375660316 Chr.18: 5394844-5394844 on Build GRCh38
rs755261359 Chr.18: 5394827-5394827 on Build GRCh38
rs965437320 Chr.18: 5394795-5394795 on Build GRCh38
rs756924326 Chr.18: 5394840-5394840 on Build GRCh38
rs1018412026 Chr.18: 5394788-5394788 on Build GRCh38
rs373966644 Chr.18: 5394670-5394670 on Build GRCh38
rs779940084 Chr.18: 5394752-5394753 on Build GRCh38
rs770926856 Chr.18: 5394637-5394637 on Build GRCh38
rs746565220 Chr.18: 5394722-5394722 on Build GRCh38
rs761758415 Chr.18: 5394683-5394683 on Build GRCh38
rs780460761 Chr.18: 5394691-5394691 on Build GRCh38
rs199616331 Chr.18: 5394687-5394687 on Build GRCh38
rs371494173 Chr.18: 5394725-5394725 on Build GRCh38
rs201592629 Chr.18: 5394783-5394783 on Build GRCh38
rs556500315 Chr.18: 5394821-5394831 on Build GRCh38
rs769674990 Chr.18: 5394841-5394841 on Build GRCh38
rs775465251 Chr.18: 5394769-5394769 on Build GRCh38
rs761207833 Chr.18: 5394800-5394800 on Build GRCh38
rs941242190 Chr.18: 5394731-5394731 on Build GRCh38
rs369448100 Chr.18: 5394635-5394635 on Build GRCh38
rs566732815 Chr.18: 5394661-5394661 on Build GRCh38
rs775413787 Chr.18: 5394667-5394667 on Build GRCh38
rs781206421 Chr.18: 5394730-5394730 on Build GRCh38
rs768953196 Chr.18: 5394790-5394790 on Build GRCh38
rs769851322 Chr.18: 5394733-5394733 on Build GRCh38
rs190875358 Chr.18: 5394753-5394753 on Build GRCh38
rs372606751 Chr.18: 5394835-5394835 on Build GRCh38
rs754220644 Chr.18: 5394821-5394821 on Build GRCh38
rs746462713 Chr.18: 5394677-5394677 on Build GRCh38
rs763335878 Chr.18: 5394668-5394668 on Build GRCh38
rs766846252 Chr.18: 5394802-5394802 on Build GRCh38
rs140830660 Chr.18: 5394686-5394686 on Build GRCh38
rs763816285 Chr.18: 5394830-5394830 on Build GRCh38
rs774369492 Chr.18: 5394791-5394791 on Build GRCh38

View Full Product Details