Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66933473-66933734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM96B
Entrez Gene ID
51647
Gene Name
family with sequence similarity 96 member B
Gene Aliases
CGI-128, MIP18
UniGene
Hs.9825
Gene Chromosome Location
Chr.16: 66932055-66934423 on Build GRCh38
Transcripts
NM_016062.3 NR_024525.2 NR_046109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427879 c.345C>T p.F115F Chr.16: 66933617-66933617 on Build GRCh38
7367560 c.348G>T p.K116N Chr.16: 66933614-66933614 on Build GRCh38
6566262 c.312G>T p.K104N Chr.16: 66933650-66933650 on Build GRCh38
4683311 c.248C>A p.A83D Chr.16: 66933714-66933714 on Build GRCh38
2993947 c.242C>T p.T81I Chr.16: 66933720-66933720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751204168 Chr.16: 66933656-66933656 on Build GRCh38
rs1054678990 Chr.16: 66933594-66933594 on Build GRCh38
rs769345145 Chr.16: 66933634-66933634 on Build GRCh38
rs750654946 Chr.16: 66933529-66933529 on Build GRCh38
rs200831328 Chr.16: 66933671-66933671 on Build GRCh38
rs755168252 Chr.16: 66933708-66933708 on Build GRCh38
rs1051297699 Chr.16: 66933670-66933670 on Build GRCh38
rs989697908 Chr.16: 66933686-66933686 on Build GRCh38
rs12811 Chr.16: 66933644-66933644 on Build GRCh38
rs991533200 Chr.16: 66933552-66933552 on Build GRCh38
rs771758179 Chr.16: 66933593-66933593 on Build GRCh38
rs967025860 Chr.16: 66933695-66933695 on Build GRCh38
rs868841066 Chr.16: 66933709-66933709 on Build GRCh38
rs762520085 Chr.16: 66933537-66933537 on Build GRCh38
rs775414472 Chr.16: 66933512-66933512 on Build GRCh38
rs781566549 Chr.16: 66933566-66933566 on Build GRCh38
rs775094816 Chr.16: 66933638-66933638 on Build GRCh38
rs750487198 Chr.16: 66933571-66933571 on Build GRCh38
rs758998063 Chr.16: 66933621-66933621 on Build GRCh38
rs915905793 Chr.16: 66933563-66933563 on Build GRCh38
rs554222442 Chr.16: 66933579-66933579 on Build GRCh38
rs746793263 Chr.16: 66933605-66933605 on Build GRCh38
rs754097771 Chr.16: 66933704-66933704 on Build GRCh38
rs764355457 Chr.16: 66933655-66933655 on Build GRCh38
rs981364455 Chr.16: 66933569-66933569 on Build GRCh38
rs374659607 Chr.16: 66933590-66933590 on Build GRCh38
rs776582844 Chr.16: 66933611-66933611 on Build GRCh38
rs780182175 Chr.16: 66933697-66933697 on Build GRCh38
rs1017035853 Chr.16: 66933527-66933527 on Build GRCh38
rs932951298 Chr.16: 66933632-66933632 on Build GRCh38
rs11556910 Chr.16: 66933690-66933690 on Build GRCh38
rs768015329 Chr.16: 66933661-66933661 on Build GRCh38
rs779007005 Chr.16: 66933711-66933711 on Build GRCh38
rs762736611 Chr.16: 66933651-66933651 on Build GRCh38
rs368141724 Chr.16: 66933602-66933602 on Build GRCh38
rs745597552 Chr.16: 66933582-66933582 on Build GRCh38
rs756229754 Chr.16: 66933687-66933687 on Build GRCh38
rs762093458 Chr.16: 66933657-66933657 on Build GRCh38
rs756280931 Chr.16: 66933580-66933580 on Build GRCh38
rs770834701 Chr.16: 66933608-66933608 on Build GRCh38
rs138252988 Chr.16: 66933541-66933541 on Build GRCh38

View Full Product Details