Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 162067909-162068182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG2
Entrez Gene ID
2566
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases
CAE2, ECA2, GEFSP3
UniGene
Hs.7195
Gene Chromosome Location
Chr.5: 162067642-162155539 on Build GRCh38
Transcripts
NM_000816.3 NM_198903.2 NM_198904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5723805 c.59C>T p.S20L Chr.5: 162068058-162068058 on Build GRCh38
3702766 c.78G>T p.W26C Chr.5: 162068077-162068077 on Build GRCh38
1253001 c.26C>G p.T9R Chr.5: 162068025-162068025 on Build GRCh38
5060899 c.107G>A p.G36D Chr.5: 162068106-162068106 on Build GRCh38
7262608 c.54_55insT p.S20fs*20 Chr.5: 162068053-162068054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375295110 Chr.5: 162068010-162068010 on Build GRCh38
rs747787220 Chr.5: 162068003-162068003 on Build GRCh38
rs769362559 Chr.5: 162068006-162068006 on Build GRCh38
rs115976622 Chr.5: 162068089-162068089 on Build GRCh38
rs754584418 Chr.5: 162067991-162067991 on Build GRCh38
rs954184248 Chr.5: 162068104-162068104 on Build GRCh38
rs770641887 Chr.5: 162068121-162068121 on Build GRCh38
rs759392289 Chr.5: 162068016-162068016 on Build GRCh38
rs771282908 Chr.5: 162067984-162067985 on Build GRCh38
rs758944561 Chr.5: 162067983-162067983 on Build GRCh38
rs780572044 Chr.5: 162067999-162067999 on Build GRCh38
rs368162707 Chr.5: 162068098-162068098 on Build GRCh38
rs777736322 Chr.5: 162067985-162067986 on Build GRCh38
rs796052502 Chr.5: 162068036-162068036 on Build GRCh38
rs765592275 Chr.5: 162067975-162067975 on Build GRCh38
rs766990192 Chr.5: 162068078-162068078 on Build GRCh38
rs61750979 Chr.5: 162068040-162068040 on Build GRCh38
rs564088820 Chr.5: 162068077-162068077 on Build GRCh38
rs367762747 Chr.5: 162067956-162067956 on Build GRCh38
rs746505697 Chr.5: 162067990-162067990 on Build GRCh38
rs943553394 Chr.5: 162068137-162068137 on Build GRCh38
rs886060381 Chr.5: 162067996-162067997 on Build GRCh38
rs764172866 Chr.5: 162068020-162068020 on Build GRCh38
rs901824125 Chr.5: 162067965-162067965 on Build GRCh38
rs972860897 Chr.5: 162068138-162068138 on Build GRCh38
rs763272278 Chr.5: 162068156-162068156 on Build GRCh38
rs750851406 Chr.5: 162067978-162067978 on Build GRCh38
rs541552445 Chr.5: 162067947-162067947 on Build GRCh38
rs796052517 Chr.5: 162068109-162068109 on Build GRCh38
rs886060382 Chr.5: 162067996-162067996 on Build GRCh38
rs780201998 Chr.5: 162067985-162067985 on Build GRCh38
rs758313419 Chr.5: 162067983-162067983 on Build GRCh38
rs770106632 Chr.5: 162068087-162068089 on Build GRCh38
rs866056788 Chr.5: 162068105-162068105 on Build GRCh38
rs183259247 Chr.5: 162068023-162068023 on Build GRCh38
rs369898285 Chr.5: 162068116-162068116 on Build GRCh38
rs376576632 Chr.5: 162068117-162068117 on Build GRCh38
rs764214358 Chr.5: 162067954-162067954 on Build GRCh38
rs780757403 Chr.5: 162067987-162067987 on Build GRCh38
rs770128365 Chr.5: 162068152-162068152 on Build GRCh38
rs55716248 Chr.5: 162068008-162068008 on Build GRCh38
rs561851806 Chr.5: 162068154-162068154 on Build GRCh38
rs771938111 Chr.5: 162068148-162068148 on Build GRCh38
rs757437605 Chr.5: 162067971-162067971 on Build GRCh38
rs776744057 Chr.5: 162068149-162068149 on Build GRCh38
rs778689358 Chr.5: 162068134-162068134 on Build GRCh38
rs750798004 Chr.5: 162068043-162068043 on Build GRCh38
rs868452487 Chr.5: 162068112-162068112 on Build GRCh38
rs961531407 Chr.5: 162068127-162068127 on Build GRCh38
rs762058667 Chr.5: 162068035-162068035 on Build GRCh38
rs780713471 Chr.5: 162068101-162068101 on Build GRCh38
rs371508835 Chr.5: 162067961-162067961 on Build GRCh38
rs781628762 Chr.5: 162067985-162067987 on Build GRCh38
rs141106898 Chr.5: 162068051-162068051 on Build GRCh38
rs199857078 Chr.5: 162068141-162068141 on Build GRCh38
rs796052516 Chr.5: 162068069-162068069 on Build GRCh38
rs36112889 Chr.5: 162067978-162067979 on Build GRCh38
rs997531721 Chr.5: 162067976-162067976 on Build GRCh38
rs774337016 Chr.5: 162068012-162068012 on Build GRCh38
rs747649928 Chr.5: 162068102-162068102 on Build GRCh38
rs200838780 Chr.5: 162068053-162068053 on Build GRCh38
rs369466385 Chr.5: 162068118-162068118 on Build GRCh38

View Full Product Details