Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172200521-172200790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB9
Entrez Gene ID
285588
Gene Name
EF-hand calcium binding domain 9
Gene Aliases
-
UniGene
Hs.716824
Gene Chromosome Location
Chr.5: 172188061-172203454 on Build GRCh38
Transcripts
NM_001171183.1 XM_017009350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403084 c.353G>A p.G118E Chr.5: 172200633-172200633 on Build GRCh38
7216368 c.417G>A p.Q139Q Chr.5: 172200697-172200697 on Build GRCh38
6251236 c.355G>T p.D119Y Chr.5: 172200635-172200635 on Build GRCh38
5918631 c.382G>A p.E128K Chr.5: 172200662-172200662 on Build GRCh38
5897523 c.367G>A p.G123S Chr.5: 172200647-172200647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759760888 Chr.5: 172200627-172200627 on Build GRCh38
rs917638708 Chr.5: 172200565-172200565 on Build GRCh38
rs77598074 Chr.5: 172200691-172200691 on Build GRCh38
rs867066300 Chr.5: 172200602-172200602 on Build GRCh38
rs372002793 Chr.5: 172200725-172200725 on Build GRCh38
rs776700402 Chr.5: 172200601-172200601 on Build GRCh38
rs1008265496 Chr.5: 172200735-172200735 on Build GRCh38
rs150351437 Chr.5: 172200603-172200603 on Build GRCh38
rs17074425 Chr.5: 172200697-172200697 on Build GRCh38
rs559270773 Chr.5: 172200748-172200748 on Build GRCh38
rs574974364 Chr.5: 172200561-172200561 on Build GRCh38
rs200613862 Chr.5: 172200645-172200645 on Build GRCh38
rs371496854 Chr.5: 172200648-172200648 on Build GRCh38
rs779288514 Chr.5: 172200707-172200707 on Build GRCh38
rs768660153 Chr.5: 172200593-172200593 on Build GRCh38
rs761912076 Chr.5: 172200605-172200605 on Build GRCh38
rs55813110 Chr.5: 172200710-172200710 on Build GRCh38
rs373569772 Chr.5: 172200655-172200655 on Build GRCh38
rs567745882 Chr.5: 172200562-172200562 on Build GRCh38
rs891172675 Chr.5: 172200716-172200716 on Build GRCh38
rs2306959 Chr.5: 172200769-172200769 on Build GRCh38
rs765056912 Chr.5: 172200616-172200616 on Build GRCh38
rs757812818 Chr.5: 172200699-172200699 on Build GRCh38
rs779590326 Chr.5: 172200745-172200745 on Build GRCh38
rs201907450 Chr.5: 172200625-172200625 on Build GRCh38
rs554892277 Chr.5: 172200559-172200559 on Build GRCh38
rs1025555860 Chr.5: 172200771-172200771 on Build GRCh38
rs746249846 Chr.5: 172200723-172200723 on Build GRCh38
rs749968228 Chr.5: 172200570-172200570 on Build GRCh38
rs922034256 Chr.5: 172200594-172200594 on Build GRCh38
rs1038613696 Chr.5: 172200686-172200686 on Build GRCh38
rs760270558 Chr.5: 172200582-172200582 on Build GRCh38
rs759658658 Chr.5: 172200548-172200548 on Build GRCh38
rs756172250 Chr.5: 172200689-172200689 on Build GRCh38
rs983754394 Chr.5: 172200563-172200563 on Build GRCh38
rs1039714880 Chr.5: 172200763-172200763 on Build GRCh38
rs902161328 Chr.5: 172200662-172200662 on Build GRCh38

View Full Product Details