Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132861541-132861802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF9
Entrez Gene ID
2661
Gene Name
growth differentiation factor 9
Gene Aliases
-
UniGene
Hs.25022
Gene Chromosome Location
Chr.5: 132861181-132866884 on Build GRCh38
Transcripts
NM_001288824.2 NM_001288825.2 NM_001288826.2 NM_001288827.2 NM_001288828.2 NM_005260.4 XM_005271957.4 XM_006714585.3 XM_011543308.2 XM_011543309.1 XM_011543310.1 XM_011543311.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3137642 c.1361G>A p.R454H Chr.5: 132861593-132861593 on Build GRCh38
1433085 c.1326G>A p.E442E Chr.5: 132861628-132861628 on Build GRCh38
5555435 c.1171G>T p.G391* Chr.5: 132861783-132861783 on Build GRCh38
4525473 c.1333G>A p.D445N Chr.5: 132861621-132861621 on Build GRCh38
3850092 c.1300C>T p.P434S Chr.5: 132861654-132861654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762144743 Chr.5: 132861604-132861604 on Build GRCh38
rs781668021 Chr.5: 132861672-132861672 on Build GRCh38
rs118080183 Chr.5: 132861671-132861671 on Build GRCh38
rs749533371 Chr.5: 132861617-132861617 on Build GRCh38
rs763654195 Chr.5: 132861749-132861749 on Build GRCh38
rs549040205 Chr.5: 132861601-132861601 on Build GRCh38
rs779083274 Chr.5: 132861703-132861703 on Build GRCh38
rs779279480 Chr.5: 132861777-132861777 on Build GRCh38
rs1002188417 Chr.5: 132861753-132861753 on Build GRCh38
rs146290291 Chr.5: 132861712-132861712 on Build GRCh38
rs756490110 Chr.5: 132861696-132861696 on Build GRCh38
rs61754582 Chr.5: 132861594-132861594 on Build GRCh38
rs753217200 Chr.5: 132861574-132861574 on Build GRCh38
rs140742945 Chr.5: 132861704-132861704 on Build GRCh38
rs369407153 Chr.5: 132861715-132861715 on Build GRCh38
rs774616473 Chr.5: 132861621-132861621 on Build GRCh38
rs116926261 Chr.5: 132861679-132861679 on Build GRCh38
rs754180892 Chr.5: 132861775-132861775 on Build GRCh38
rs983773241 Chr.5: 132861663-132861663 on Build GRCh38
rs755189977 Chr.5: 132861776-132861776 on Build GRCh38
rs754503543 Chr.5: 132861576-132861576 on Build GRCh38
rs909663056 Chr.5: 132861773-132861773 on Build GRCh38
rs759188524 Chr.5: 132861723-132861723 on Build GRCh38
rs142443414 Chr.5: 132861654-132861654 on Build GRCh38
rs768761463 Chr.5: 132861618-132861618 on Build GRCh38
rs764883262 Chr.5: 132861734-132861734 on Build GRCh38
rs373342775 Chr.5: 132861716-132861716 on Build GRCh38
rs761806089 Chr.5: 132861624-132861624 on Build GRCh38
rs374221648 Chr.5: 132861709-132861709 on Build GRCh38
rs748451406 Chr.5: 132861781-132861781 on Build GRCh38
rs762446912 Chr.5: 132861748-132861748 on Build GRCh38
rs766066211 Chr.5: 132861570-132861570 on Build GRCh38
rs191882278 Chr.5: 132861625-132861625 on Build GRCh38
rs376173165 Chr.5: 132861772-132861772 on Build GRCh38
rs771874845 Chr.5: 132861758-132861762 on Build GRCh38
rs752226758 Chr.5: 132861656-132861656 on Build GRCh38
rs377177196 Chr.5: 132861650-132861650 on Build GRCh38
rs767055173 Chr.5: 132861700-132861700 on Build GRCh38
rs748661153 Chr.5: 132861590-132861591 on Build GRCh38
rs976692105 Chr.5: 132861635-132861635 on Build GRCh38
rs775778786 Chr.5: 132861612-132861612 on Build GRCh38
rs756623311 Chr.5: 132861757-132861757 on Build GRCh38
rs34172592 Chr.5: 132861750-132861750 on Build GRCh38
rs200924295 Chr.5: 132861631-132861631 on Build GRCh38
rs893869185 Chr.5: 132861763-132861763 on Build GRCh38
rs141444900 Chr.5: 132861652-132861652 on Build GRCh38
rs932305992 Chr.5: 132861660-132861660 on Build GRCh38
rs146595130 Chr.5: 132861651-132861651 on Build GRCh38
rs372761965 Chr.5: 132861642-132861642 on Build GRCh38
rs778344377 Chr.5: 132861577-132861577 on Build GRCh38
rs188623863 Chr.5: 132861593-132861593 on Build GRCh38
rs369636792 Chr.5: 132861698-132861698 on Build GRCh38

View Full Product Details