Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172659862-172660135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASLG
Entrez Gene ID
356
Gene Name
Fas ligand
Gene Aliases
ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6, TNLG1A
UniGene
Hs.2007
Gene Chromosome Location
Chr.1: 172659008-172666873 on Build GRCh38
Transcripts
NM_000639.2 NM_001302746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5574619 c.373T>G p.S125A Chr.1: 172660119-172660119 on Build GRCh38
5378439 c.378T>A p.S126S Chr.1: 172660124-172660124 on Build GRCh38
2075323 c.378T>G p.S126S Chr.1: 172660124-172660124 on Build GRCh38
7367621 c.364C>A p.H122N Chr.1: 172660110-172660110 on Build GRCh38
899689 c.382G>A p.E128K Chr.1: 172660128-172660128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372615226 Chr.1: 172660106-172660106 on Build GRCh38
rs887612686 Chr.1: 172660028-172660028 on Build GRCh38
rs778330321 Chr.1: 172660050-172660050 on Build GRCh38
rs936149515 Chr.1: 172659898-172659898 on Build GRCh38
rs751740630 Chr.1: 172659936-172659936 on Build GRCh38
rs978013524 Chr.1: 172660042-172660042 on Build GRCh38
rs751847640 Chr.1: 172660112-172660112 on Build GRCh38
rs114673102 Chr.1: 172659928-172659928 on Build GRCh38
rs889414217 Chr.1: 172659914-172659914 on Build GRCh38
rs975550035 Chr.1: 172659972-172659972 on Build GRCh38
rs371453329 Chr.1: 172660017-172660017 on Build GRCh38
rs557574091 Chr.1: 172660022-172660022 on Build GRCh38
rs762222796 Chr.1: 172660113-172660113 on Build GRCh38
rs747781769 Chr.1: 172660062-172660062 on Build GRCh38
rs541791449 Chr.1: 172659956-172659956 on Build GRCh38
rs770652587 Chr.1: 172660082-172660082 on Build GRCh38
rs760081633 Chr.1: 172659975-172659977 on Build GRCh38
rs151243922 Chr.1: 172660027-172660027 on Build GRCh38
rs750918300 Chr.1: 172660045-172660045 on Build GRCh38
rs370125821 Chr.1: 172660071-172660071 on Build GRCh38
rs140406512 Chr.1: 172660110-172660110 on Build GRCh38
rs142254412 Chr.1: 172659990-172659990 on Build GRCh38
rs754474862 Chr.1: 172660048-172660048 on Build GRCh38
rs367759983 Chr.1: 172659896-172659896 on Build GRCh38
rs759431087 Chr.1: 172660097-172660097 on Build GRCh38
rs897465194 Chr.1: 172659954-172659954 on Build GRCh38
rs776590086 Chr.1: 172660092-172660092 on Build GRCh38
rs746745833 Chr.1: 172660079-172660079 on Build GRCh38
rs771774232 Chr.1: 172660066-172660066 on Build GRCh38
rs550196951 Chr.1: 172659900-172659900 on Build GRCh38

View Full Product Details