Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35107677-35107912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM214B
Entrez Gene ID
80256
Gene Name
family with sequence similarity 214 member B
Gene Aliases
KIAA1539
UniGene
Hs.301696
Gene Chromosome Location
Chr.9: 35104121-35116350 on Build GRCh38
Transcripts
NM_001317991.1 NM_025182.3 NR_134455.1 XM_005251588.1 XM_005251590.1 XM_005251591.1 XM_005251592.1 XM_005251593.1 XM_005251594.1 XM_005251596.4 XM_005251597.4 XM_005251598.4 XM_011518037.1 XM_011518039.1 XM_011518040.1 XM_011518043.1 XM_011518044.1 XM_017015169.1 XM_017015170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308454 c.502C>T p.Q168* Chr.9: 35107773-35107773 on Build GRCh38
6659475 c.440delC p.P147fs*50 Chr.9: 35107835-35107835 on Build GRCh38
7367326 c.532G>A p.A178T Chr.9: 35107743-35107743 on Build GRCh38
3952688 c.514G>T p.G172W Chr.9: 35107761-35107761 on Build GRCh38
6895680 c.509C>T p.S170F Chr.9: 35107766-35107766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756522517 Chr.9: 35107840-35107840 on Build GRCh38
rs747808726 Chr.9: 35107871-35107871 on Build GRCh38
rs776223436 Chr.9: 35107764-35107764 on Build GRCh38
rs752963471 Chr.9: 35107834-35107834 on Build GRCh38
rs759840013 Chr.9: 35107712-35107712 on Build GRCh38
rs781394793 Chr.9: 35107721-35107721 on Build GRCh38
rs13287211 Chr.9: 35107768-35107768 on Build GRCh38
rs764414755 Chr.9: 35107841-35107841 on Build GRCh38
rs757819380 Chr.9: 35107846-35107846 on Build GRCh38
rs7039828 Chr.9: 35107861-35107861 on Build GRCh38
rs10972317 Chr.9: 35107759-35107759 on Build GRCh38
rs751796669 Chr.9: 35107714-35107714 on Build GRCh38
rs773264963 Chr.9: 35107783-35107783 on Build GRCh38
rs751743252 Chr.9: 35107819-35107819 on Build GRCh38
rs375238152 Chr.9: 35107825-35107825 on Build GRCh38
rs771547340 Chr.9: 35107751-35107751 on Build GRCh38
rs779607163 Chr.9: 35107752-35107752 on Build GRCh38
rs758424101 Chr.9: 35107857-35107857 on Build GRCh38
rs773205168 Chr.9: 35107889-35107889 on Build GRCh38
rs753115560 Chr.9: 35107725-35107725 on Build GRCh38
rs766559637 Chr.9: 35107808-35107808 on Build GRCh38
rs746529007 Chr.9: 35107848-35107848 on Build GRCh38
rs778373655 Chr.9: 35107739-35107739 on Build GRCh38
rs61735988 Chr.9: 35107743-35107743 on Build GRCh38
rs754310832 Chr.9: 35107842-35107842 on Build GRCh38
rs77226241 Chr.9: 35107891-35107891 on Build GRCh38
rs779482350 Chr.9: 35107847-35107847 on Build GRCh38
rs769603526 Chr.9: 35107886-35107886 on Build GRCh38
rs754548616 Chr.9: 35107859-35107859 on Build GRCh38
rs763027871 Chr.9: 35107793-35107793 on Build GRCh38
rs150618944 Chr.9: 35107716-35107716 on Build GRCh38
rs558758005 Chr.9: 35107729-35107729 on Build GRCh38
rs61735989 Chr.9: 35107769-35107769 on Build GRCh38
rs756510814 Chr.9: 35107732-35107732 on Build GRCh38
rs183501601 Chr.9: 35107770-35107770 on Build GRCh38
rs746651794 Chr.9: 35107758-35107758 on Build GRCh38
rs767781512 Chr.9: 35107829-35107829 on Build GRCh38

View Full Product Details