Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37459505-37459764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR1
Entrez Gene ID
23070
Gene Name
cap methyltransferase 1
Gene Aliases
FTSJD2, KIAA0082, MTr1, hMTr1
UniGene
Hs.520102
Gene Chromosome Location
Chr.6: 37433131-37482844 on Build GRCh38
Transcripts
NM_015050.2 XM_005248955.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3718781 c.1067_1068insG p.V358fs*7 Chr.6: 37459656-37459657 on Build GRCh38
1078674 c.1013G>A p.R338H Chr.6: 37459602-37459602 on Build GRCh38
5482745 c.982G>A p.G328S Chr.6: 37459571-37459571 on Build GRCh38
6667675 c.1089T>C p.A363A Chr.6: 37459678-37459678 on Build GRCh38
3076577 c.1063C>T p.R355C Chr.6: 37459652-37459652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200782942 Chr.6: 37459718-37459718 on Build GRCh38
rs949907974 Chr.6: 37459578-37459578 on Build GRCh38
rs780356385 Chr.6: 37459698-37459698 on Build GRCh38
rs199815717 Chr.6: 37459602-37459602 on Build GRCh38
rs530421853 Chr.6: 37459696-37459696 on Build GRCh38
rs751159738 Chr.6: 37459666-37459666 on Build GRCh38
rs376267984 Chr.6: 37459559-37459559 on Build GRCh38
rs748752404 Chr.6: 37459711-37459711 on Build GRCh38
rs755018501 Chr.6: 37459530-37459530 on Build GRCh38
rs755416126 Chr.6: 37459705-37459705 on Build GRCh38
rs561385958 Chr.6: 37459642-37459642 on Build GRCh38
rs772267740 Chr.6: 37459615-37459615 on Build GRCh38
rs769179894 Chr.6: 37459604-37459604 on Build GRCh38
rs777409300 Chr.6: 37459663-37459663 on Build GRCh38
rs746177112 Chr.6: 37459599-37459599 on Build GRCh38
rs770140756 Chr.6: 37459601-37459601 on Build GRCh38
rs752195687 Chr.6: 37459653-37459653 on Build GRCh38
rs138183916 Chr.6: 37459709-37459709 on Build GRCh38
rs377098871 Chr.6: 37459652-37459652 on Build GRCh38
rs773298348 Chr.6: 37459616-37459616 on Build GRCh38
rs996591746 Chr.6: 37459659-37459659 on Build GRCh38
rs753730251 Chr.6: 37459626-37459626 on Build GRCh38
rs368232854 Chr.6: 37459703-37459703 on Build GRCh38
rs149591587 Chr.6: 37459712-37459712 on Build GRCh38
rs778537640 Chr.6: 37459535-37459535 on Build GRCh38
rs757852955 Chr.6: 37459654-37459654 on Build GRCh38
rs1054057486 Chr.6: 37459648-37459648 on Build GRCh38
rs766339236 Chr.6: 37459549-37459550 on Build GRCh38
rs546430261 Chr.6: 37459714-37459714 on Build GRCh38
rs771311761 Chr.6: 37459720-37459720 on Build GRCh38
rs764308181 Chr.6: 37459643-37459643 on Build GRCh38
rs774439790 Chr.6: 37459609-37459609 on Build GRCh38
rs1004771511 Chr.6: 37459710-37459710 on Build GRCh38
rs368591877 Chr.6: 37459613-37459613 on Build GRCh38
rs183699274 Chr.6: 37459545-37459545 on Build GRCh38
rs776521307 Chr.6: 37459733-37459733 on Build GRCh38
rs959453740 Chr.6: 37459538-37459538 on Build GRCh38
rs935902565 Chr.6: 37459647-37459647 on Build GRCh38
rs372628932 Chr.6: 37459625-37459625 on Build GRCh38
rs777305793 Chr.6: 37459568-37459568 on Build GRCh38

View Full Product Details