Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119156423-119156696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG4
Entrez Gene ID
64137
Gene Name
ATP binding cassette subfamily G member 4
Gene Aliases
WHITE2
UniGene
Hs.126378
Gene Chromosome Location
Chr.11: 119149040-119162666 on Build GRCh38
Transcripts
NM_001142505.1 NM_022169.4 XM_011542952.2 XM_017018121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7121580 c.832C>G p.Q278E Chr.11: 119156585-119156585 on Build GRCh38
6537536 c.815A>G p.Y272C Chr.11: 119156568-119156568 on Build GRCh38
4658476 c.851T>C p.V284A Chr.11: 119156604-119156604 on Build GRCh38
5371803 c.880G>A p.G294R Chr.11: 119156633-119156633 on Build GRCh38
337235 c.900C>T p.P300P Chr.11: 119156653-119156653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770520477 Chr.11: 119156667-119156667 on Build GRCh38
rs529515398 Chr.11: 119156551-119156551 on Build GRCh38
rs781633638 Chr.11: 119156490-119156490 on Build GRCh38
rs376368836 Chr.11: 119156561-119156561 on Build GRCh38
rs1002065770 Chr.11: 119156571-119156571 on Build GRCh38
rs777812747 Chr.11: 119156504-119156504 on Build GRCh38
rs746879394 Chr.11: 119156610-119156610 on Build GRCh38
rs755846434 Chr.11: 119156453-119156456 on Build GRCh38
rs781038169 Chr.11: 119156632-119156632 on Build GRCh38
rs746980624 Chr.11: 119156534-119156534 on Build GRCh38
rs754126746 Chr.11: 119156605-119156605 on Build GRCh38
rs779072546 Chr.11: 119156609-119156609 on Build GRCh38
rs752353129 Chr.11: 119156622-119156622 on Build GRCh38
rs760114542 Chr.11: 119156572-119156572 on Build GRCh38
rs776518922 Chr.11: 119156541-119156541 on Build GRCh38
rs755234776 Chr.11: 119156480-119156480 on Build GRCh38
rs936896707 Chr.11: 119156518-119156518 on Build GRCh38
rs778011563 Chr.11: 119156526-119156526 on Build GRCh38
rs749146911 Chr.11: 119156529-119156529 on Build GRCh38
rs986593942 Chr.11: 119156598-119156598 on Build GRCh38
rs562419971 Chr.11: 119156546-119156546 on Build GRCh38
rs372377217 Chr.11: 119156529-119156529 on Build GRCh38
rs571407889 Chr.11: 119156536-119156536 on Build GRCh38
rs753076717 Chr.11: 119156600-119156600 on Build GRCh38
rs368360612 Chr.11: 119156467-119156467 on Build GRCh38
rs766975905 Chr.11: 119156460-119156460 on Build GRCh38
rs375589685 Chr.11: 119156549-119156549 on Build GRCh38
rs369406470 Chr.11: 119156639-119156639 on Build GRCh38
rs757261121 Chr.11: 119156608-119156608 on Build GRCh38
rs771070110 Chr.11: 119156667-119156668 on Build GRCh38
rs764288490 Chr.11: 119156603-119156603 on Build GRCh38
rs748966571 Chr.11: 119156654-119156654 on Build GRCh38
rs769546546 Chr.11: 119156547-119156547 on Build GRCh38
rs774666899 Chr.11: 119156570-119156570 on Build GRCh38
rs369798507 Chr.11: 119156638-119156638 on Build GRCh38
rs756362410 Chr.11: 119156525-119156525 on Build GRCh38
rs754911669 Chr.11: 119156625-119156625 on Build GRCh38
rs749348580 Chr.11: 119156528-119156528 on Build GRCh38
rs371672140 Chr.11: 119156559-119156559 on Build GRCh38
rs752816611 Chr.11: 119156519-119156519 on Build GRCh38
rs368442729 Chr.11: 119156555-119156555 on Build GRCh38
rs150494059 Chr.11: 119156599-119156599 on Build GRCh38
rs756521751 Chr.11: 119156602-119156602 on Build GRCh38
rs201199426 Chr.11: 119156668-119156668 on Build GRCh38
rs772929341 Chr.11: 119156650-119156650 on Build GRCh38

View Full Product Details